Skip to Content
Merck
All Photos(3)

Key Documents

HPA020459

Sigma-Aldrich

Anti-KIAA0391 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MRPP3, Anti-PRORP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SYVSLFLADRCGIRNQQRLFSLKTMSPQNTKATNLIAKARYLRKDEGSNKQVYSVPHFFLAGAAKERSQMNSQTEDHALAPVRNTIQLPTQPLNSEEWDKLKEDLKENTGKTSFESWIISQMAGCHSSIDVAKSLLAW

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPP3(9692)

General description

Mitochondrial ribonuclease P protein 3 (MRPP3) is a subunit of mitochondrial-RNase P complex, with RNA-binding and metallonuclease domains. It belongs to the pentatricopeptide repeat (PPR) domain protein family. The gene encoding this protein is localized on human chromosome 14q13.2.

Immunogen

Mitochondrial ribonuclease P protein 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-MRPP3 Prestige Antibodies® Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Mitochondrial ribonuclease P protein 3 (MRPP3) influences the expression of RNAs with the help of other proteins. It is also required for the cleavage of the 5′ tRNA ends effectively.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70556

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of the nuclease subunit of human mitochondrial RNase P.
Reinhard L, et al.
Nucleic Acids Research, 43(11), 5664-5672 (2015)
Scott C Walker et al.
Cell, 135(3), 412-414 (2008-11-06)
In bacteria, archaea, and the eukaryote nucleus, the endonuclease ribonuclease P (RNase P) is composed of a catalytic RNA that is assisted by protein subunits. Holzmann et al. (2008) now provide evidence that the human mitochondrial RNase P is an
Oliver Rackham et al.
RNA (New York, N.Y.), 17(12), 2085-2093 (2011-10-27)
Human mitochondrial long noncoding RNAs (lncRNAs) have not been described to date. By analysis of deep-sequencing data we have identified three lncRNAs generated from the mitochondrial genome and confirmed their expression by Northern blotting and strand-specific qRT-PCR. We show that
Maria I G Lopez Sanchez et al.
Molecular endocrinology (Baltimore, Md.), 29(1), 14-27 (2014-11-07)
Estrogens, in particular 17β-estradiol, are well-known regulators of essential cellular functions; however, discrepancies remain over the mechanisms by which they act on mitochondria. Here we propose a novel mechanism for the direct regulation of mitochondrial gene expression by estrogen under
Osama Alsmadi et al.
Annals of human genetics, 73(Pt 5), 475-483 (2009-07-25)
The role of the KIAA0391 and PSMA6 genes in predisposing individuals to disease is still not fully understood. We evaluated by molecular beacon-based genotyping assays the roles of five single nucleotide polymorphisms (SNPs) in the chromosomal region 14q13.2 harbouring the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service