Skip to Content
Merck
All Photos(8)

Key Documents

MX647S100

Sigma-Aldrich

Mix-n-Stain CF 647 Antibody Labeling Kit (50-100μg)

Synonym(s):

Antibody Labeling Kit, CF 647 Labeling Kit, Mix-n-Stain Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

form

liquid

storage condition

protect from light

fluorescence

λex 650 nm; λem 665 nm

storage temp.

−20°C

General description

This kit contains everything you need to rapidly label an antibody. The labeling procedure comprises simple mixing of your antibody and the optimally formulated dye in the reaction buffer provided, followed by a brief incubation. The resulting solution is ready for staining without further purification. The kit is suitable for labeling commercially available primary antibodies, either directly or after a simple antibody clean-up step. After labeling, the dye is covalently linked to the antibody.
This labeling kit is for labeling 50-100 μg antibody with the far red fluorescent dye CF647. Mix-n-Stain CF dye antibody labeling kits are a revolutionary antibody labeling technology that permits one to label antibodies with our superior CF dyes in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. CF647 is far-red fluorescent dye and one of an outstanding series of CF dyes. CF647 is spectrally similar to Alexa Fluor 647 and Cy®5 dyes. CF dyes are superior to Alexa Fluor dyes and Cy® dyes for antibody labeling by having combined advantages in brightness, photostability, specificity and novel features ideal for in vivo imaging.

Application

The Mix-n-Stain CF dye antibody labeling kit can be used to label primary antibodies for ELISA, western blot, immunohistochemistry, flow cytometry, FACS, immunofluorescence or immunocytochemistry applications.

Features and Benefits

A major advantage of the labeling kits is the superior CF dyes, many of which are brighter and more photostable than Alexa Fluor dyes and other similar commercial dyes. With a large list of CF dyes of wavelengths covering the entire visible and near-infrared spectra, you have unprecedented freedom to label any commercial primary antibody with a color of your choice, making multi-color detection simple and affordable.
Mix-n-Stain CF dye antibody labeling kits dramatically simplify the process of preparing fluorescently labeled antibodies, particularly primary antibodies. Simply mix your antibody with a CF dye of your choice in the buffer provided, a step that takes less than 30 seconds of hands-on time. After waiting for another 30 minutes, without a separation step, you will have a fluorescent antibody conjugate that is as good as the best pre-labeled fluorescent antibody you can buy from some of the leading suppliers. There is no need to calculate how much dye you should use; just mix your antibody with the entire amount of dye provided and you will always produce optimal labeling. Moreover, the labeling reaction can tolerate the presence of common stabilizers, such as sodium azide, Tris, glycerol and low level of BSA.

Components

The kit consists of 1 vial each of Mix-n-Stain Dye, Mix-n-Stain Reaction Buffer (10x), Mix-n-Stain Storage Buffer, and an ultrafiltration vial.

Analysis Note

Direct replacement for: Alexa Fluor 647, Cy®5, DY- 647, DyLight® 649, IRDye® 650

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Cy is a registered trademark of Cytiva
DyLight is a registered trademark of Pierce Biotechnology, Inc.
IRDye is a registered trademark of LI-COR, Inc.
Mix-n-Stain is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marion Pardons et al.
Cell reports, 29(9), 2783-2795 (2019-11-28)
Latent proviruses persist in central (TCM), transitional (TTM), and effector (TEM) memory cells. We measured the levels of cellular factors involved in HIV gene expression in these subsets. The highest levels of acetylated H4, active nuclear factor κB (NF-κB), and
Identification of a mesenchymal progenitor cell hierarchy in adipose tissue
Merrick D, et al.
Science, 364(6438) (2019)
Latency-Reversing Agents Induce Differential Responses in Distinct Memory CD4 T Cell Subsets in Individuals on Antiretroviral Therapy
Pardons M, et al.
Cell Reports, 29(9), 2783-2795 (2019)
Irfan Ullah et al.
bioRxiv : the preprint server for biology (2021-04-02)
Neutralizing antibodies (NAbs) are effective in treating COVID-19 but the mechanism of immune protection is not fully understood. Here, we applied live bioluminescence imaging (BLI) to monitor the real-time effects of NAb treatment in prophylaxis and therapy of K18-hACE2 mice
David Merrick et al.
Science (New York, N.Y.), 364(6438) (2019-04-27)
Metabolic health depends on the capacity of adipose tissue progenitor cells to undergo de novo adipogenesis. The cellular hierarchy and mechanisms governing adipocyte progenitor differentiation are incompletely understood. Through single-cell RNA sequence analyses, we show that the lineage hierarchy of

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service