Skip to Content
Merck
All Photos(5)

Key Documents

HPA018531

Sigma-Aldrich

Anti-FOS antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AP-1, Anti-c-fos

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DLQWLVQPALVSSVAPSQTRAPHPFGVPAPSAGAYSRAGVVKTMTGGRAQSIGRRGKVEQLSPEEEEKRRIRRERNKMAAAKCRNRRRELTDTLQAETDQLEDEKSALQTEIANLLKEKEKLEFILAAHRPACKIPDDL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOS(2353)

General description

The gene C-FOS (Cellular oncogene fos) is mapped to human chromosome 14q24.3. It belongs to AP-1 (activator protein-1) family of transcription factors. The protein is mainly localized in the nucleus.

Immunogen

Proto-oncogene protein c-fos recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-FOS antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

C-FOS (Cellular oncogene fos) is considered immediate-early gene. Its expression is generally low but rapidly increases in response to extracellular stimuli that result in cellular processes such as cell proliferation and differentiation. C-FOS forms AP-1 (activator protein-1) heterodimer complex with C-JUN (Proto-oncogene c-Jun) and thereby acts as a transcriptional regulator. C-FOS regulates genes which are important for tumorigenesis. Change in expression of C-FOS is associated with psychosis-proneness or schizotypy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70428

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chu-Chu Qi et al.
Behavioral and brain functions : BBF, 14(1), 7-7 (2018-03-21)
Following a social defeat, the balanced establishment and extinction of aversive information is a beneficial strategy for individual survival. Abnormal establishment or extinction is implicated in the development of mental disorders. This study investigated the time course of the establishment
K Okazaki et al.
The EMBO journal, 14(20), 5048-5059 (1995-10-16)
The c-mos proto-oncogene product, Mos, is a serine/threonine kinase that can activate ERK1 and 2 mitogen-activated protein (MAP) kinases by direct phosphorylation of MAPK/ERK kinase (MEK). ERK activation is essential for oncogenic transformation of NIH 3T3 cells by Mos. In
P Grant et al.
European psychiatry : the journal of the Association of European Psychiatrists, 29(5), 304-306 (2014-03-19)
Psychosis-proneness or schizotypy is a personality organisation mirroring individual risk for schizophrenia-development. Believed to be a fully dimensional construct sharing considerable geno- and phenotypal variance with clinical schizophrenia, it has become an increasingly promising tool for basic psychosis-research. Although many
L L Bonnycastle et al.
Neuroscience letters, 160(1), 33-36 (1993-09-17)
A gene for early-onset familial Alzheimer's disease (FAD) is located on chromosome 14q24.3. The c-fos gene (FOS) is also located in the same band of this chromosome and is thus a candidate for the FAD locus. A yeast artificial chromosome
M M Portal et al.
Oncogene, 26(24), 3551-3558 (2006-12-13)
c-Fos dephosphorylated on tyrosine (c-Fos), a component of the activator protein-1 (AP-1) family of transcription factors, is expressed at very low levels in resting cells. However, its expression is rapidly upregulated in cells undergoing G(0) to S phase transition leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service