Skip to Content
Merck
All Photos(7)

Key Documents

HPA019149

Sigma-Aldrich

Anti-JUNB antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Transcription factor jun-B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SLHDYKLLKPSLAVNLADPYRSLKAPGARGPGPEGGGGGSYFSGQGSDTGASLKLASSELERLIVPNSNGVITTTPTPPGQYFYPRGGGSGGGAGGAGGGVTEEQEGFADGFVKALDDL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JUNB(3726)

General description

The gene JUNB (Jun B proto-oncogene) is mapped to human chromosome 19p13.2. It belongs to activator protein 1 (AP-1) family of transcription factors. JUNB is ubiquitously expressed.

Immunogen

Transcription factor jun-B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

JUNB (Jun B proto-oncogene) is a transcription factor. It is up-regulated in primary placental mesenchymal stromal cells (PDMSCs) obtained from preeclamptic (PE) pregnancies and affects cyclin-D1 regulation. A2B (adenosine receptor)-mediated up-regulation of JUNB regulates VEGF (vascular endothelial growth factor) generation and angiogenesis. During acute hepatitis, JUNB controls interferon (IFN)-γ production in NK (natural killer) and NK-T cells, and thereby induces cell death. It is over-expressed in Hodgkin lymphoma, CD30+ diffuse large B cell lymphoma and anaplastic large cell lymphoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84804

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason K H Lee et al.
The Journal of biological chemistry, 288(30), 21482-21495 (2013-06-12)
The activator protein-1 (AP-1) family transcription factor, JunB, is an important regulator of proliferation, apoptosis, differentiation, and the immune response. In this report, we show that JunB is cleaved in a caspase-dependent manner in apoptotic anaplastic lymphoma kinase-positive, anaplastic large
Sergey Ryzhov et al.
Molecular pharmacology, 85(1), 62-73 (2013-10-19)
Interstitial adenosine stimulates neovascularization in part through A2B adenosine receptor-dependent upregulation of vascular endothelial growth factor (VEGF). In the current study, we tested the hypothesis that A2B receptors upregulate JunB, which can contribute to stimulation of VEGF production. Using the
Siri Sæterstad et al.
PloS one, 17(3), e0265189-e0265189 (2022-03-12)
In recent years it has become apparent that the epithelium is highly involved in inflammatory bowel disease (IBD) pathophysiology. The majority of gene expression studies of IBD are generated from heterogeneous biopsies, providing no distinction between immune cells, the epithelium
A M Nuzzo et al.
Placenta, 35(7), 483-490 (2014-05-02)
In the present study, we characterized the expression of Activating Protein 1 (AP-1) factors, key cell cycle regulators, in primary placental mesenchymal stromal cells (PDMSCs) derived from normal and preeclamptic (PE) pregnancies with fetal-placental compromise. PDMSCs were isolated from control
J Spring
FEBS letters, 400(1), 2-8 (1997-01-02)
For the growing fraction of human genes with identified functions there are often homologues known from invertebrates such as Drosophila. A survey of well established gene families from aldolases to zinc finger transcription factors reveals that usually a single invertebrate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service