Skip to Content
Merck
All Photos(4)

Key Documents

HPA008791

Sigma-Aldrich

Anti-MYBL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A-Myb antibody produced in rabbit, Anti-Myb-related protein A antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

EANAVLSSLQTIPEFAETLELIESDPVAWSDVTSFDISDAAASPIKSTPVKLMRIQHNEGAMECQFNVSLVLEGKKNTCNGGNSEAVPLTSPNIAKFSTPPAILRKKRKMRVGHSPGSELRDGSLNDGGN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYBL1(4603)

Related Categories

General description

MYBL1 (v-myb avian myeloblastosis viral oncogene homolog-like 1) gene is localized to human chromosome 8q13.1, and codes for a transcription factor. It is a member of the oncoprotein family called Myb. Its N-terminal contains DNA-binding repeats, and its C-terminal contains two conserved regions. Its c-Myb transactivation domain contains a putative leucine zipper.

Immunogen

Myb-related protein A recombinant protein epitope signature tag (PrEST)

Application

Anti-MYBL1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MYBL1 (v-myb avian myeloblastosis viral oncogene homolog-like 1) is partially duplicated in 28% of diffuse astrocytoma grade II tumors. This gene is characterized by recurrent oncogenic truncating rearrangements in pediatric low-grade gliomas. This gene is thought to be involved in the control of differentiation and proliferation of normal B-cells and Burkitt′s lymphoma cells. It also acts as an interacting partner of CBP (CREB-binding protein) protein, where its charged sequence binds with the CREB (cAMP response element-binding protein)-binding domain of CBP.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71002

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshitsugu Mitani et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(3), 725-733 (2015-12-04)
Adenoid cystic carcinoma (ACC) is an indolent salivary gland malignancy, characterized by t(6;9) translocations and MYB-NFIB gene fusions in approximately 50% of the tumors. The genetic alterations underlying t(6;9)-negative and t(6;9)-positive/MYB-NFIB fusion-negative ACC remain unknown. To uncover the genetic alterations
Robin Kohler et al.
Nucleic acids research, 52(12), 6830-6849 (2024-05-15)
A-MYB (MYBL1) is a transcription factor with a role in meiosis in spermatocytes. The related B-MYB protein is a key oncogene and a master regulator activating late cell cycle genes. To activate genes, B-MYB forms a complex with MuvB and
Lori A Ramkissoon et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(20), 8188-8193 (2013-05-02)
Pediatric low-grade gliomas (PLGGs) are among the most common solid tumors in children but, apart from BRAF kinase mutations or duplications in specific subclasses, few genetic driver events are known. Diffuse PLGGs comprise a set of uncommon subtypes that exhibit
V Facchinetti et al.
The Biochemical journal, 324 ( Pt 3), 729-736 (1997-06-15)
The A-Myb transcription factor belongs to the Myb family of oncoproteins and is likely to be involved in the regulation of proliferation and/or differentiation of normal B cells and Burkitt's lymphoma cells. To characterize in detail the domains of A-Myb
Brooke E Howitt et al.
The Journal of pathology, 235(1), 37-49 (2014-09-19)
Müllerian adenosarcoma (MA) is a rare mixed mesenchymal tumour of the female genital tract, composed of malignant stroma and benign-appearing epithelium. Sarcomatous overgrowth (SO) is the only established histological variable associated with higher stage and shorter survival. Specific molecular or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service