Skip to Content
Merck
All Photos(1)

Key Documents

342M-1

Sigma-Aldrich

Tryptase (G3) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

diluted ascites fluid

antibody product type

primary antibodies

clone

G3, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (342M-14)
vial of 0.5 mL concentrate (342M-15)
bottle of 1.0 mL predilute (342M-17)
vial of 1.0 mL concentrate (342M-16)
bottle of 7.0 mL predilute (342M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200-1:1000

isotype

IgG1

control

uterus

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... TPSAB1(7177)

General description

Tryptases constitute a subfamily of trypsin-like proteinases, stored in the mast cell secretory granules and basophils. Upon cellular activation, these enzymes are released into the extracellular environment. Anti-tryptase is a good marker for mast cells, basophils, and their derivatives.

Quality


IVD

IVD

IVD

RUO

Linkage

Tryptase Positive Control Slides, Product No. 342S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Y Li
Leukemia research, 25(7), 537-541 (2001-05-30)
The diagnosis of mastocytosis or mast cell disease may be difficult sometimes because of the wide variety of clinical presentation, abnormal morphology of mast cells, and variation in histologic features which may mimic varieties of other diseases. Over the years
L K Gordon et al.
Clinical immunology (Orlando, Fla.), 94(1), 42-50 (1999-12-23)
pANCA is a marker antibody expressed in most patients with ulcerative colitis, and its cognate antigen is potentially an immunologic target in this disease. This study evaluates whether pANCA detects an autoantigen that is expressed in the colonic mucosa. Immunohistochemistry
J H Jordan et al.
Human pathology, 32(5), 545-552 (2001-06-19)
In an attempt to identify novel diagnostic markers for mast cell (MC)-proliferative disorders, serial bone marrow (bm) sections of 22 patients with mastocytosis (systemic indolent mastocytosis, n = 19; mast cell leukemia [MCL], n = 1; isolated bm mastocytosis, n
Andreas Chott et al.
The American journal of surgical pathology, 27(7), 1013-1019 (2003-06-27)
Mast cell sarcoma is an extremely rare and aggressive type of mast cell disease. Only a few cases have been described so far, and little is known about the biology and phenotype of afflicted cells. We describe morphologic and immunophenotypic
L Fiorucci et al.
Cellular and molecular life sciences : CMLS, 61(11), 1278-1295 (2004-06-02)
Tryptases constitute a subfamily of trypsin-like proteinases, stored in the mast cell secretory granules of all mammalian organisms. These enzymes are released along with other mediators into the extracellular medium upon mast cell activation/degranulation. Among the trypsin-like enzymes, tryptases are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service