Skip to Content
Merck
All Photos(1)

Key Documents

HPA004879

Sigma-Aldrich

Anti-SCN7A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Putative voltage-gated sodium channel α subunit Nax antibody produced in rabbit, Anti-Sodium channel protein type 7 subunit α antibody produced in rabbit, Anti-Sodium channel protein type VII subunit α antibody produced in rabbit, Anti-Sodium channel protein, cardiac and skeletal muscle α-subunit antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

LLKILCKTQNVPKDTMDHVNEVYVKEDISDHTLSELSNTQDFLKDKEKSSGTEKNATENESQSLIPSPSVSETVPIASGESDIENLDNKEIQSKSGDGGSKEK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCN7A(6332)

General description

SCN7A (sodium channel, voltage-gated, type VII, alpha subunit) is a large subunit (260kDa) of heteromultimeric voltage-gated sodium channel belonging to the voltage-dependent sodium channel family. In humans, it is expressed in the brain, fetal skeletal muscle, heart, and uterus. It is associated with membrane depolarization of electrically excitable cells, including other diverse physiological processes.

Immunogen

Sodium channel protein type 7 subunit α recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86836

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Bogdanovic et al.
IUBMB life, 72(4), 601-606 (2020-02-07)
The sodium channel NaX (encoded by the SCN7A gene) was originally identified in the heart and skeletal muscle and is structurally similar to the other voltage-gated sodium channels but does not appear to be voltage gated. Although NaX is expressed
A L George et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(11), 4893-4897 (1992-06-11)
Previously cloned voltage-dependent sodium channels exhibit a high degree of homology to one another and appear to comprise a single multigene family. We have now isolated and characterized cDNAs from both human adult heart and fetal skeletal muscle that encode
Kirsi Penttinen et al.
BMC cardiovascular disorders, 23(1), 208-208 (2023-04-26)
Brugada syndrome is an inherited cardiac arrhythmia disorder that is mainly associated with mutations of the cardiac voltage-gated sodium channel alpha subunit 5 (SCN5A) gene. The clinical symptoms include ventricular fibrillation and an increased risk of sudden cardiac death. Human-induced
Ping Liang et al.
Journal of the American College of Cardiology, 68(19), 2086-2096 (2016-11-05)
Brugada syndrome (BrS), a disorder associated with characteristic electrocardiogram precordial ST-segment elevation, predisposes afflicted patients to ventricular fibrillation and sudden cardiac death. Despite marked achievements in outlining the organ level pathophysiology of the disorder, the understanding of human cellular phenotype
Assignment of a human voltage-dependent sodium channel alpha-subunit gene (SCN6A) to 2q21-q23.
A L George et al.
Genomics, 19(2), 395-397 (1994-01-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service