Skip to Content
Merck
All Photos(2)

Documents

C2687

Sigma-Aldrich

Monoclonal Anti-Calponin antibody produced in mouse

clone hCP, ascites fluid

Synonym(s):

Anti-Calponin-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

hCP, monoclonal

mol wt

antigen 34 kDa

contains

15 mM sodium azide

species reactivity

rat, rabbit, pig, human, mouse

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1:10,000 using human uterus extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

The antibody hCP (also cited as CALP), localizes calponin in mammalian smooth muscle. In human uterus, an additional band of approximately 27 kDa (l-calponin) may also be stained. The antibody does not cross-react with skeletal, cardiac or non-muscle tissue calponin. Nevertheless, the antibody exhibits cross-reactivity with an epitope (150 kDa range) in human or mouse skeletal muscle. In immunohistochemical staining, the product exhibits smooth muscle specificity. It stains vascular and visceral smooth muscle cells in tissue sections and primary cultured cells (or early passages), but not most cell lines originally derived from smooth muscle. The antibody does not stain epithelial, endothelial or connective tissue fibroblast cells. This product does not cross react with smooth muscle tissue from chicken.

Immunogen

human uterus smooth muscle extract.

Application

Monoclonal Anti-Calponin antibody produced in mouse is suitable for the following applications:
  • Immunohistochemistry using formalin-fixed, paraffin-embedded sections
  • Immunoprecipitation
  • Microarray
  • Western blotting (at a dilution of 1:10,000 using human uterus extract)
  • Flow cytometry analysis

Biochem/physiol Actions

Calponin is a calcium binding protein that belongs to a family of actin-associated proteins. Calponin is necessary for autophosphorylation of protein kinase C (PKC) in vascular smooth muscle (VSM). Calponin-1 inhibition can prevent uterine smooth muscle cell migration, cause morphological change and rearrange F-actin without affecting its proliferation and apoptosis. Calponin h1 (CN) is a differentiation marker of smooth muscle cells and is down-regulated in the blood vessels of several human tumors. It can inhibit actomyosin ATPase activity. The h1 and h2 calponins bind F-actin and play a key role in regulating actin filaments in smooth muscle and non-muscle cells.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mun Chun Chan et al.
Molecular and cellular biology, 31(3), 517-530 (2010-12-08)
Pulmonary artery hypertension (PAH) is characterized by elevated pulmonary artery resistance and increased medial thickness due to deregulation of vascular remodeling. Inactivating mutations of the BMPRII gene, which encodes a receptor for bone morphogenetic proteins (BMPs), are identified in ∼60%
Gaëlle Pérot et al.
Cancer research, 69(6), 2269-2278 (2009-03-12)
Myocardin (MYOCD), a serum response factor (SRF) transcriptional cofactor, is essential for cardiac and smooth muscle development and differentiation. We show here by array-based comparative genomic hybridization, fluorescence in situ hybridization, and expression analysis approaches that MYOCD gene is highly
Y Yanagisawa et al.
European journal of surgical oncology : the journal of the European Society of Surgical Oncology and the British Association of Surgical Oncology, 34(5), 531-537 (2007-08-21)
Calponin h1 (CN) is a differentiation marker of smooth muscle cells that has been reported to be down-regulated in the blood vessels of several human tumors. In this study, we examined CN expression in blood vessels in relation to the
Kris M Mann et al.
The Journal of cell biology, 165(4), 483-491 (2004-05-19)
The process of vascular smooth muscle cell (vSMC) differentiation is critical to embryonic angiogenesis. However, despite its importance, the vSMC differentiation program remains largely undefined. Murine gene disruption studies have identified several gene products that are necessary for vSMC differentiation
Yong-hong Gu et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 30(6), 1369-1372 (2010-06-30)
To study the effects of calponin-1 expression inhibition on the proliferation , invasiveness, apoptosis and cytoskeleton of uterine smooth muscle cells, and explore the molecular mechanism of calponin-1 in the uterine smooth muscle cells for labor onset. siRNA-calponin-1 adenovirus plasmid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service