Skip to Content
Merck
All Photos(1)

Key Documents

H6916

Sigma-Aldrich

Macrophage Colony-Stimulating Factor human

≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Synonym(s):

MCSF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Macrophage Colony-Stimulating Factor human, M-CSF, recombinant, expressed in HEK 293 cells, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

≤4 ng/mL EC50

quality

endotoxin tested

mol wt

dimer 35-40 kDa (glycosylated)

packaging

pkg of 10 μg

manufacturer/tradename

HumanZyme

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CSF1(1435)

Looking for similar products? Visit Product Comparison Guide

General description

M-CSF is encoded by the gene mapped to human chromosome 1p13.3. It is a glycosylated polypeptide, which exists as a homodimer. M-CSF is produced by macrophage, fibroblasts, lymphocyte and endothelial cells.

Biochem/physiol Actions

Macrophage CSF (M-CSF) as a growth factor, stimulates the proliferation and differentiation of monocyte/macrophage-lineage cells. It also boosts antibody-dependent, cell-mediated cytotoxicity by monocytes and macrophages. In addition, human M-CSF inhibits bone resorption by osteoclasts. It might also play a crucial role in atherosclerosis. Overexpression of the gene has been observed in Paget′s disease of bone (PDB).

Physical form

Lyophilized from a 0.2 μm filtered solution of 1x PBS.

Analysis Note

The specific activity was determined by the dose-dependent stimulation of the proliferation of murine M-NFS-60 cells (mouse myeloid Leukemia indicator cell line).

Citation

1. Mazur, E.M., and Cohen, J.L., Clin. Pharmacol. Ther., 46, 250 (1989).
2. Morstyn, G., and Burgess, A.W., Cancer Res., 48, 5624 (1988).
3. Metcalf, D., Blood, 67, 257 (1986).
4. Mufson, R.A., et al., Cell. Immunol., 119, 182 (1989).
5. Hattersley, G., et al., J. Cell Physiol., 137, 199 (1988).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association study identifies variants at CSF1, OPTN and TNFRSF11A as genetic risk factors for Paget's disease of bone.
Albagha O M E, et al.
Nature Genetics, 429(6), 520-520 (2010)
Macrophage colony-stimulating factor enhances monocyte and macrophage antibody-dependent cell-mediated cytotoxicity.
Mufson R A, et al.
Cellular Immunology, 119(1), 182-192 (1989)
Human macrophage colony-stimulating factor inhibits bone resorption by osteoclasts disaggregated from rat bone.
Hattersley G, et al.
Journal of Cellular Physiology, 137(1), 199-203 (1988)
The molecular biology and functions of the granulocyte-macrophage colony-stimulating factors.
Metcalf D.
Blood, 67(2), 257-267 (1986)
"Effects of macrophage colony-stimulating factor (M-CSF) on protease production from monocyte, macrophage and foam cell in vitro: a possible mechanism for anti-atherosclerotic effect of M-CSF.
Tojo N, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1452.3, 275-284 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service