Skip to Content
Merck
All Photos(3)

Key Documents

H6515

Sigma-Aldrich

L-Homoserine

Synonym(s):

(S)-2-Amino-4-hydroxybutyric acid, Hse

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721681
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

203 °C (dec.) (lit.)

application(s)

cell analysis
detection

SMILES string

N[C@@H](CCO)C(O)=O

InChI

1S/C4H9NO3/c5-3(1-2-6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1

InChI key

UKAUYVFTDYCKQA-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Homoserine is a variant of serine with an additional carbon on its side chain.

Application

L-Homoserine has been used as an internal standard for neurotransmitter analysis and amino acids quantification.

Biochem/physiol Actions

L-Homoserine is synthesized by deoxidation process, catalysed by homoserine dehydrogenase. This is one of the steps in the synthesis of L-threonine. The carbon flux in in bacteria such as E. coli is maintained by this reaction.
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chiral recognition of non-natural ?-amino acids
Gal JF, et al.
International Journal of Mass Spectrometry, 222(1?3), 259-267 (2003)
Reprogramming Microbial Metabolic Pathways, 288-288 (2012)
Neurotoxic effect of maneb in rats as studied by neurochemical and immunohistochemical parameters
Nielsen BS, et al.
Environmental Toxicology and Pharmacology, 21(3), 268-275 (2006)
Minsang Lee et al.
The Plant journal : for cell and molecular biology, 41(5), 685-696 (2005-02-11)
Homoserine kinase (HSK) produces O-phospho-l-homoserine (HserP) used by cystathionine gamma-synthase (CGS) for Met synthesis and threonine synthase (TS) for Thr synthesis. The effects of overexpressing Arabidopsis thaliana HSK, CGS, and Escherichia coli TS (eTS), each controlled by the 35S promoter
Jorge E Vidal et al.
Infection and immunity, 81(4), 1341-1353 (2013-02-14)
Streptococcus pneumoniae forms organized biofilms in the human upper respiratory tract that may play an essential role in both persistence and acute respiratory infection. However, the production and regulation of biofilms on human cells is not yet fully understood. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service