Skip to Content
Merck
All Photos(2)

Key Documents

H4628

Sigma-Aldrich

DL-Homocysteine

≥95% (titration)

Synonym(s):

2-Amino-4-mercaptobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
135.18
Beilstein:
1721683
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
42021308
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Homocysteine, ≥95% (titration)

Assay

≥95% (titration)

form

powder

mp

232-233 °C (lit.)

solubility

H2O: soluble

application(s)

cell analysis
peptide synthesis

storage temp.

−20°C

SMILES string

NC(CCS)C(O)=O

InChI

1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)

InChI key

FFFHZYDWPBMWHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Homocysteine has been used to induce hyperhomocysteinemia in Sprague–Dawley rats. It has also been used to study the effects of hyperhomocysteinemia on atherosclerosis in apolipoprotein E-deficient mice.

Biochem/physiol Actions

Homocysteine is a sulfhydryl-containing amino acid, synthesized from methionine. It is a non-essential, non-proteinogenic amino acid. It is an important determinant of the methylation cycle and is present in the plasma in four forms. An abnormally high level of homocysteine leads to hyperhomocysteinemia and also promotes atherosclerosis.
Increased plasma homocysteine is a risk factor for coronary heart disease and carcinogenesis; may be a marker for tumor cell growth.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyperhomocysteinemia decreases bone blood flow
Neetu Tyagi
Vascular Health, 7, 31-35 (2011)
Homocysteine: role and implications in atherosclerosis.
Guthikonda S and Haynes WG
Current Atherosclerosis Reports, 8(2), 100-106 (2006)
The Molecular and Cellular Effect of Homocysteine Metabolism Imbalance on Human Health
Henrieta S
International Journal of Molecular Sciences, 17(10), 1733-1733 (2016)
Massimo Terzolo et al.
The Journal of clinical endocrinology and metabolism, 89(8), 3745-3751 (2004-08-05)
We evaluated serum homocysteine concentrations and the C677T polymorphism of the gene encoding for methylene tetrahydrofolate reductase, a key enzyme for homocysteine metabolism, in 57 patients with Cushing's syndrome, 41 with active disease, and 16 in remission after successful surgery
Wolfgang C Winkelmayer et al.
Journal of the American Society of Nephrology : JASN, 16(1), 255-260 (2004-11-26)
Homocysteine is implicated to be an atherogenic amino acid and has been associated with increased risk of adverse cardiovascular outcomes. The prognostic significance of plasma total homocysteine (tHcy) levels for mortality and allograft loss in kidney transplant recipients has not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service