Skip to Content
Merck
All Photos(1)

Key Documents

H3537

Sigma-Aldrich

HEPES solution

99.5%, liquid, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered

Synonym(s):

N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Product Name

HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered

grade

BioPerformance Certified

Quality Level

sterility

0.2 μm filtered

form

liquid

concentration

1 M

technique(s)

cell culture | mammalian: suitable

impurities

Bioburden, tested
DNase, RNase, Protease, Nickase, free
endotoxin, tested

pH

5.0-6.0

useful pH range

6.8-8.2

cation traces

Fe: <5 ppm
heavy metals (as Pb): <5 ppm

suitability

suitable for molecular biology

application(s)

diagnostic assay manufacturing

SMILES string

OCCN1CCN(CCS(=O)(O)=O)CC1

InChI

1S/C8H18N2O4S/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14/h11H,1-8H2,(H,12,13,14)

InChI key

JKMHFZQWWAIEOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

HEPES has been described as one of the best all-purpose buffers available for biological research.[1] At biological pH, the molecule is zwitterionic, and is effective as a buffer at pH 6.8 to 8.2. HEPES has been used in a wide variety of applications, including tissue culture. It is commonly used to buffer cell culture media in air. HEPES finds its usage in in vitro experiments on Mg.[2]

Application

RNAse HEPES has been used:
  • To supplement Dulbecco′smodified Eagle′smedium for maintenance of cell line and RPMI medium to wash rat islets[3]
  • To recover purified ribonucleotide[4]
  • As a component of HEPES/KOH buffer and adenylation buffer for small RNA isolation and sequencing[5]
  • As a component in buffers used for nuclear extract preparation and also to supplement RPMI-1640 medium for maintenance of islets[6]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Small RNA Isolation and Sequencing Protocol and Its Application to Assay CRISPR RNA Biogenesis in Bacteria.
Silas S, et al.
Bio-protocol, 8(4) (2018)
Timing of CRISPR/Cas9-related mRNA microinjection after activation as an important factor affecting genome editing efficiency in porcine oocytes.
Sato M, et al.
Theriogenology, 108, 29-38 (2018)
Expression of an uncleavable N-terminal RasGAP fragment in insulin-secreting cells increases their resistance toward apoptotic stimuli without affecting their glucose-induced insulin secretion.
Yang JY, et al.
The Journal of Biological Chemistry, 280(38), 32835-32842 (2005)
Masahiro Sato et al.
Theriogenology, 108, 29-38 (2017-12-02)
Recently, successful one-step genome editing by microinjection of CRISPR/Cas9-related mRNA components into the porcine zygote has been described. Given the relatively long gestational period and the high cost of housing swine, the establishment of an effective microinjection-based porcine genome editing
Masahiro Sato et al.
International journal of molecular sciences, 16(8), 17838-17856 (2015-08-08)
Some reports demonstrated successful genome editing in pigs by one-step zygote microinjection of mRNA of CRISPR/Cas9-related components. Given the relatively long gestation periods and the high cost of housing, the establishment of a single blastocyst-based assay for rapid optimization of

Articles

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service