Skip to Content
Merck
All Photos(1)

Documents

H3041

Sigma-Aldrich

Halopemide

≥98% (HPLC)

Synonym(s):

N-(2-(4-(5-Chloro-2-oxo-1-benzimidazolinyl)piperidino)ethyl)-p-fluorobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClFN4O2
CAS Number:
Molecular Weight:
416.88
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)NCCN2CCC(CC2)N3C(=O)Nc4cc(Cl)ccc34

InChI

1S/C21H22ClFN4O2/c22-15-3-6-19-18(13-15)25-21(29)27(19)17-7-10-26(11-8-17)12-9-24-20(28)14-1-4-16(23)5-2-14/h1-6,13,17H,7-12H2,(H,24,28)(H,25,29)

InChI key

NBHPRWLFLUBAIE-UHFFFAOYSA-N

Application

Halopemide, a non-specific phospholipase D (PLD) antagonist, may be used with selective PLD antagonists (CAY10593, a PLD1 antagonist; CAY10594 or ML298, selective PLD2 antagonist) to help define the role and physiological effects regulated by phospholipase D enzymes. Halopemide may be used as the basis of the design and development of more selective PLD antagonists.

Biochem/physiol Actions

Halopemide is a dopamine receptor antagonist and a phospholipase D2 inhibitor. Halopemide may be used as a screen to identify inhibitors of human PLD2 using an in vitro biochemical assay. It is also inhibitory at benzodiazepine binding sites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A J Loonen et al.
Archives internationales de pharmacodynamie et de therapie, 247(1), 43-58 (1980-09-01)
The effects of halopemide on the release of 3H-serotonin, 3H-noradrenaline, 3H-acetylcholine and 3H-GABA from rat frontal cortical slices in vitro were studied and compared to those of its neuroleptic congener R29800, spiperone and haloperidol. Spontaneous 3H-serotonin and to a lesser
Effects of halopemide on GABA receptor binding, uptake and release.
A J Loonen et al.
Brain research, 210(1-2), 485-492 (1981-04-06)
R Neale et al.
Psychopharmacology, 75(3), 254-257 (1981-01-01)
Oxiperomide and tiapride are dopamine receptor antagonists claimed to have "antidyskinetic" properties in animal models and the clinic. Halopemide and mezilamine are other dopamine antagonists predicted to lack extrapyramidal side effects in man on the basis of animal studies. Acute
A J Loonen et al.
Archives internationales de pharmacodynamie et de therapie, 258(1), 51-59 (1982-07-01)
Halopemide inhibits 3H-BZ binding to crude and washed rat forebrain membranes with IC50 values of 16-25 microM. Its putative metabolites are considerably less active. The actions of halopemide are probably not directly related to its ability to interfere with high-affinity
H H van Rooij et al.
Journal of chromatography, 164(2), 177-185 (1979-10-11)
Dynamic (solvent generated) cation-exchange systems for the separation of drugs and main metabolites derived from butyrophenone and diphenylpiperidine (haloperidol, pimozide, halopemide) were investigated. The effect of organic modifier, detergent, counter-ion concentration and of the pH on the retention has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service