Skip to Content
Merck
All Photos(1)

Key Documents

B157

Sigma-Aldrich

BQ-788

≥95%, solid

Synonym(s):

2,6-Dimethylpiperidinecarbonyl-γ-Methyl-Leu-Nin-(Methoxycarbonyl)-D-Trp-D-Nle, N-[N-[N-[(2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl]-1-(methoxycarbonyl)-D-tryptophyl]-D-norleucine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H50N5NaO7
CAS Number:
Molecular Weight:
663.78
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥95%

form

solid

color

white

solubility

acetonitrile: 0.3 mg/mL
DMSO: 1.2 mg/mL
ethanol: 1.2 mg/mL
H2O: slightly soluble

storage temp.

−20°C

SMILES string

[Na+].CCCC[C@@H](NC(=O)[C@@H](Cc1cn(C(=O)OC)c2ccccc12)NC(=O)[C@H](CC(C)(C)C)NC(=O)N3[C@@H](C)CCC[C@H]3C)C([O-])=O

InChI

1S/C34H51N5O7.Na/c1-8-9-16-25(31(42)43)35-29(40)26(18-23-20-38(33(45)46-7)28-17-11-10-15-24(23)28)36-30(41)27(19-34(4,5)6)37-32(44)39-21(2)13-12-14-22(39)3;/h10-11,15,17,20-22,25-27H,8-9,12-14,16,18-19H2,1-7H3,(H,35,40)(H,36,41)(H,37,44)(H,42,43);/q;+1/p-1/t21-,22+,25-,26-,27+;/m1./s1

InChI key

QCVIFBRTTLMEOV-FUKQNADPSA-M

Gene Information

human ... EDNRB(1910)
mouse ... EDNRB(13618)
rat ... EDNRB(50672)

Looking for similar products? Visit Product Comparison Guide

Application

BQ-788 has been used as a selective endothelin-B (ETRB) blocker in human airway smooth muscle cells (HASMCs), uterine mesothelial cells (UtMCs)-derived vascular smooth muscle cells (VSMCs), and human umbilical vein endothelial cells (HUVECs)

Biochem/physiol Actions

Selective ETB endothelin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Lenyta Oliveira Gomes et al.
The Journal of pharmacy and pharmacology, 70(7), 893-900 (2018-03-24)
This study assessed the ability of endothelin-1 (ET-1) to evoke heat hyperalgesia when injected directly into the trigeminal ganglia (TG) of mice and determined the receptors implicated in this effect. The effects of TG ETA and ETB receptor blockade on
Christophe Faisy et al.
Critical care (London, England), 15(5), R208-R208 (2011-09-15)
Lung inflation may have deleterious effects on the alveoli during mechanical ventilation. However, the consequences of stretch during excessive lung inflation on basal tone and responsiveness of human bronchi are unknown. This study was undertaken to devise an experimental model
Zhuo Wang et al.
PloS one, 8(3), e58078-e58078 (2013-04-05)
Obstructive sleep apnoea (OSA) is a risk factor for cardiovascular disorders and in some cases is complication of pulmonary hypertension. We simulated OSA by exposing rats to cyclic intermittent hypoxia (CIH) to investigate its effect on pulmonary vascular endothelial dysfunction.
Emilia Kansanen et al.
Molecular pharmacology, 92(4), 481-490 (2017-08-06)
Nitro-fatty acids are reactive signaling mediators that are formed when unsaturated fatty acids react with nitric oxide or nitric oxide-derived species. Nitro-fatty acids can modify specific signaling pathways via post-translational modifications of Cys residues in key regulatory proteins. One of
Megan M Wenner et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 313(1), R51-R57 (2017-04-26)
Endothelin-1 (ET-1) contributes to age-related endothelial dysfunction in men via the ET

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service