Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥85%
storage temp.
−20°C
SMILES string
[Li+].[Li+].[Li+].O.CCCCCC(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)([O-])=O)n2cnc3c(N)ncnc23
InChI
1S/C27H46N7O17P3S.3Li.H2O/c1-4-5-6-7-18(36)55-11-10-29-17(35)8-9-30-25(39)22(38)27(2,3)13-48-54(45,46)51-53(43,44)47-12-16-21(50-52(40,41)42)20(37)26(49-16)34-15-33-19-23(28)31-14-32-24(19)34;;;;/h14-16,20-22,26,37-38H,4-13H2,1-3H3,(H,29,35)(H,30,39)(H,43,44)(H,45,46)(H2,28,31,32)(H2,40,41,42);;;;1H2/q;3*+1;/p-3/t16-,20-,21-,22+,26-;;;;/m1..../s1
InChI key
YMMJFYLVSRICJX-ZTADYLHKSA-K
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| description ≥85% | description ≥90% (HPLC) | description ~90% (HPLC) | description ≥85% |
| assay ≥85% | assay ≥90% (HPLC) | assay ~90% (HPLC) | assay ≥85% |
| Quality Level 200 | Quality Level 200 | Quality Level 100 | Quality Level 200 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



