Skip to Content
Merck
All Photos(1)

Key Documents

28007

Sigma-Aldrich

Crotonoyl coenzyme A trilithium salt

~90% (HPLC)

Synonym(s):

trans-2-Butenoyl coenzyme A trilithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37Li3N7O17P3S
Molecular Weight:
853.41
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Assay:
~90% (HPLC)
form:
solid
solubility:
H2O: 50 mg/mL, clear, colorless
Pricing and availability is not currently available.

Assay

~90% (HPLC)

Quality Level

form

solid

impurities

≤10% water

solubility

H2O: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].C\C=C\C(=O)SCCNC(=O)CCNC(=O)[C@H](O)C(C)(C)COP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP(O)([O-])=O)n2cnc3c(N)ncnc23

InChI

1S/C25H40N7O17P3S.3Li/c1-4-5-16(34)53-9-8-27-15(33)6-7-28-23(37)20(36)25(2,3)11-46-52(43,44)49-51(41,42)45-10-14-19(48-50(38,39)40)18(35)24(47-14)32-13-31-17-21(26)29-12-30-22(17)32;;;/h4-5,12-14,18-20,24,35-36H,6-11H2,1-3H3,(H,27,33)(H,28,37)(H,41,42)(H,43,44)(H2,26,29,30)(H2,38,39,40);;;/q;3*+1/p-3/b5-4+;;;/t14-,18-,19-,20+,24-;;;/m1.../s1

InChI key

LUYQFKSDLNQLEG-OUWZIDFPSA-K

Application

Crotonoyl coenzyme A (Crotonoyl -CoA) may be used in anti-tubercular and anti-malarial drug research as a substrate to help identify and characterized enzymes such as Plasmodium falciparum enoyl-ACP reductase (PfENR) and other enoyl-CoA reductases that may be effective drug targets. Crotonoyl -CoA may be used as a substrate analogue for kinetic studies on ß-hydroxyacyl-acyl carrier protein (ACP) dehydratase (FabZ).

Other Notes

Oxidation-reduction of general acyl-CoA dehydrogenase by the butyryl-CoA/crotonyl-CoA couple[1]; Purification of rat liver microsomal trans-2-enoyl-CoA reductase[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peng Liao et al.
Biochemical and biophysical research communications, 524(3), 730-735 (2020-02-10)
Post-translational modifications (PTMs) play pivotal roles in controlling the stability and activity of the tumor suppressor p53 in response to distinct stressors. Here we report an unexpected finding of a short chain fatty acid modification of p53 in human cells.
A new β-hydroxyacyl-acyl carrier protein dehydratase (FabZ) from Helicobacter pylori: Molecular cloning, enzymatic characterization, and structural modeling.
Liu W, Luo C, Han C, Peng S, et al.
Biochemical and Biophysical Research Communications, 12, 1078-1086 (2005)
Purification of rat liver microsomal trans-2-enoyl-CoA reductase.
C F Chiang
Preparative biochemistry, 17(4), 315-325 (1987-01-01)
Neha Kapoor et al.
IUBMB life, 61(11), 1083-1091 (2009-10-28)
A structure-based approach has been adopted to develop 2'-substituted analogs of triclosan. The Cl at position 2' in ring B of triclosan was chemically substituted with other functional groups like NH(2), NO(2) and their inhibitory potencies against PfENR were determined.
Sara Tucci et al.
FEBS letters, 581(8), 1561-1566 (2007-03-27)
An NADH-dependent trans-2-enoyl-CoA reductase (EC1.1.1.36) from the Gram negative spirochete Treponema denticola was identified, expressed and biochemically characterized. The recombinant protein is a monomeric enzyme with a molecular mass of 44 kDa with a specific activity of 43+/-4.8 U/mg (micromol

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service