Skip to Content
Merck
All Photos(2)

Key Documents

G2505

Sigma-Aldrich

N-Glutaryl-L-phenylalanine p-nitroanilide

protease substrate

Synonym(s):

Glutaryl-L-phenylalanine 4-nitroanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21N3O6
CAS Number:
Molecular Weight:
399.40
Beilstein:
2919832
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

methanol with 1 M NH4OH: 50 mg/mL, clear to slightly hazy

storage temp.

−20°C

SMILES string

OC(=O)CCCC(=O)N[C@@H](Cc1ccccc1)C(=O)Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C20H21N3O6/c24-18(7-4-8-19(25)26)22-17(13-14-5-2-1-3-6-14)20(27)21-15-9-11-16(12-10-15)23(28)29/h1-3,5-6,9-12,17H,4,7-8,13H2,(H,21,27)(H,22,24)(H,25,26)/t17-/m0/s1

InChI key

LFZGBNATHRHOKZ-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Glutaryl-L-phenylalanine p-nitroanilide has been used as a substrate to determine chymotrypsin activity.

Biochem/physiol Actions

N-Glutaryl-L-phenylalanine p-nitroanilide is useful in testing α-chymotrypsin activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Spreti et al.
European journal of biochemistry, 268(24), 6491-6497 (2001-12-12)
alpha-Chymotrypsin activity was tested with N-glutaryl-l-phenylalanine p-nitroanilide (GPNA) in aqueous media in the presence of synthetic surfactants, which differ in the flexibility of their bulky head groups. Superactivity can be ascribed to the presence of the tributylammonium residue on the
S V Verevka
Ukrainskii biokhimicheskii zhurnal (1978), 68(3), 36-41 (1996-05-01)
Data on the kinetics of S2'-stimulated alpha-chymotrypsin action have been presented. It is supposed that the increase of the catalytic action of S2'-stimulated chymotrypsin occurs at all three stages of the hydrolytic process-at the enzyme-substrate complex formation, its transformation to
New properties of chymotrypsin modified by fixation with a hydrophobic molecule: hexanal.
M H Remy et al.
Annals of the New York Academy of Sciences, 434, 343-346 (1984-01-01)
S Blais et al.
The Journal of biological chemistry, 268(25), 18637-18639 (1993-09-05)
The Michaelis constant of alpha-chymotrypsin, immobilized on a glutaraldehyde-activated silicate support, for N-glutaryl-L-phenylalanine-p-nitroanilide was determined and was found to be identical with that of the enzyme in solution. The influence of intraparticular diffusion was taken into account by immobilizing different
Daniel Brugger et al.
The British journal of nutrition, 116(3), 425-433 (2016-05-28)
This study investigated the effects of short-term subclinical Zn deficiency on exocrine pancreatic activity and changes in digestive capacity. A total of forty-eight weaned piglets were fed ad libitum a basal diet (maize and soyabean meal) with adequate Zn supply

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service