Skip to Content
Merck
All Photos(2)

Key Documents

G10050

Sigma-Aldrich

Sephadex® G-100 Size Exclusion Resin

Superfine, 285-700 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

product name

Sephadex® G-100, Superfine

technique(s)

affinity chromatography: suitable

matrix active group

phase

swelling

1 g swells to 15-20 mL

bead size

20-50 μm

application(s)

life science and biopharma

compatibility

Cytiva

Looking for similar products? Visit Product Comparison Guide

General description

G10050-100G′s updated product number is GE17-0061-01

Application

Fractionation Range (MW)
Globular Proteins: 4,000 - 100,000
Dextrans: 1,000 - 100,000
Sephadex®G-100 is a gel filtration medium used in protein chromatography, affinity chromatography and gel filtration chromatography.

Legal Information

Sephadex is a registered trademark of Cytiva

replaced by

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mandeep Kaur et al.
Journal of immunology (Baltimore, Md. : 1950), 202(6), 1885-1894 (2019-02-03)
Development of targeted cancer therapy requires a thorough understanding of mechanisms of tumorigenesis as well as mechanisms of action of therapeutics. This is challenging because by the time patients are diagnosed with cancer, early events of tumorigenesis have already taken
Serdar Güvercin et al.
Protein and peptide letters, 15(1), 6-12 (2008-01-29)
Glutathione S-transferase was purified from bovine erythrocytes and some kinetic and characteristic properties of the enzyme were investigated. The purification procedure was composed of preparation of homogenate and Glutathione-Agarose affinity chromatography. Thanks to the procedure, the enzyme was purified 6,800
Aisha Siddiqua et al.
Biological & pharmaceutical bulletin, 31(5), 802-808 (2008-05-03)
Low molecular weight acid phosphatase (LM-ACP) peak 2 (the isoenzyme corresponding to isoform 2, IF-2) from the liver of fish Rahu (Labeo rohita) was purified to homogeneity. 900 times purification resulted with specific activity of 35 U/mg of protein and
Andre Müller et al.
The FEBS journal, 274(24), 6329-6339 (2007-11-22)
cDNA for octopine dehydrogenase (ODH) from the adductor muscle of the great scallop, Pecten maximus, was cloned using 5'- and 3'-RACE. The cDNA comprises an ORF of 1197 nucleotides and the deduced amino acid sequence encodes a protein of 399
Evan S Leibner et al.
Acta biomaterialia, 5(5), 1389-1398 (2009-01-13)
Analytical protocol greatly influences the measurement of human serum albumin (HSA) adsorption to commercial expanded polytetrafluororethylene (ePTFE) exhibiting superhydrophobic wetting properties. Degassing of buffer solutions and evacuation of ePTFE adsorbent to remove trapped air immediately prior to contact with protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service