Skip to Content
Merck
All Photos(1)

Key Documents

S6782

Sigma-Aldrich

Superdex®

Superdex®200 Prep Grade

Synonym(s):

Size exclusion media, Superdex column

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

suspension (20% aqueous ethanol)

technique(s)

protein array: suitable

matrix

cross-linked agarose

particle size

24-44 μm

pore size

1-100 kDa MW range (dextrans)
10-600 kDa MW range (proteins)

operating pH range

1-14(short term)
3-12(long term)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Combines the high selectivity of a Sephadex matrix with the chemical and physical stability of crosslinked agarose.

Application

Optimized for high resolution preparative separations by size exclusion chromatography. Recommended fractionation ranges are listed for globular proteins and dextrans.
Superdex® is used for protein chromatography, gel filtration chromatography, gel filtration media, resins and separation media. Superdex® has been used for the purification and characterization of a novel laccase from the edible mushroom Hericium coralloides. Superdex® has also been used for the purification and mechanism exploration of a potential human hepatocellular carcinoma inhibitor from Bauhinia purpurea L. seeds.

Legal Information

Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Ouellette et al.
Protein expression and purification, 30(2), 156-166 (2003-07-26)
A recombinant form of human rhIL-7 was overexpressed in Escherichia coli HMS174 (DE3) pLysS under the control of a T7 promoter. The resulting insoluble inclusion bodies were separated from cellular debris by cross-flow filtration and solubilized by homogenization with 6
Mario Jiz et al.
Infection and immunity, 76(7), 3164-3169 (2008-04-23)
Despite effective chemotherapy, schistosomiasis remains a major public health problem in the developing world, with at least 200 million active infections resulting in significant morbidity. Rapid reinfection after treatment, accompanied by extensive residual morbidity, mandates alternative control strategies, including vaccine
Purification of sucrose synthase from rice and its protein-chemical characterization.
Elling, L., and Kula, M.-R.
Journal of Biotechnology, 29(3), 277-286 (1993)
J L Van Hove et al.
Biochemistry and molecular biology international, 43(3), 613-623 (1997-11-14)
Large quantities of recombinant acid alpha-glucosidase are needed for in vivo experimentation of enzyme replacement therapy in Pompe disease. We describe a new purification method for the purification of this recombinant enzyme from tissue culture medium consisting of concanavalin A
J Oh et al.
Human molecular genetics, 9(3), 375-385 (2000-02-03)
Hermansky-Pudlak syndrome (HPS) is a rare autosomal recessive disorder in which oculocutaneous albinism, bleeding tendency and a ceroid-lipofuscin lysosomal storage disease result from defects of multiple cytoplasmic organelles: melanosomes, platelet dense granules and lysosomes. The HPS polypeptide, a 700 amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service