Skip to Content
Merck
All Photos(1)

Key Documents

G0262

Sigma-Aldrich

Gelatin hydrolysate Enzymatic

Synonym(s):

Peptone from gelatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

Porcine

Quality Level

form

powder

Looking for similar products? Visit Product Comparison Guide

General description

Gelatin hydrolysate is extracted from pigs, bovines, and fish by the hydrolysis of gelatin.

Application

Gelatin hydrolysate Enzymatic has been used :
  • in immunoassay
  • as an excipient in vaccine formulation to screen its ability to stabilize influenza vaccine
  • for the synthesis of gelatin hydrolysate vinyl esters

Biochem/physiol Actions

Gelatin hydrolysate possesses antioxidant, angiotensin-converting enzyme (ACE)-inhibitory, antifreeze, and antiphotoaging activities. It is widely used in the food and pharmaceutical industries. Gelatin hydrolysate is used as a smart drug delivery carrier for cancer therapy and in wound dressing. It boosts skin recovery by supporting wound closure and diminishing skin wrinkles. When orally consumed, gelatin hydrolysates increase bone mass and inhibit osteopenia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydrolysis Process Optimization and Functional Characterization of Yak Skin Gelatin Hydrolysates
Yang H, et al.
Journal of Chemistry (2019)
F Elsaesser
Neuroendocrinology, 74(5), 288-299 (2001-11-06)
It has repeatedly been suggested that galanin acts within the anterior pituitary (AP) in an auto/paracrine manner to modulate luteinizing hormone (LH) release. Except for one recent report in the rat, evidence for this notion is absent. The purpose of
Min-Hsuan Lin et al.
Microbiology spectrum, 10(6), e0394922-e0394922 (2022-11-30)
More than 100 arboviruses, almost all of which have an RNA genome, cause disease in humans. RNA viruses are causing unprecedented health system challenges worldwide, many with little or no specific therapies or vaccines available. Certain species of mosquito can
Development of a stable liquid formulation of live attenuated influenza vaccine
White JA, et al.
Vaccine, 34(32), 3676-3683 (2016)
Three-dimensional microfabrication of protein hydrogels via two-photon-excited thiol-vinyl ester photopolymerization
Qin XH, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 51(22), 4799-4810 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service