Skip to Content
Merck
All Photos(1)

Key Documents

920029

Sigma-Aldrich

Low endotoxin non-gelling gelatin from porcine skin

Mw ≤6500

Synonym(s):

X-PURE® 10HGP

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

mol wt

Mw ≤6500

impurities

≤10 EU/g Endotoxin

color

white to off-white

storage temp.

2-8°C

Application

This low endotoxin gelatin is a non-gelling gelatin extracted from porcine skin suitable for vaccine and injectable formulations. Also, gelatin can be functionalized with crosslinkable groups. Non-gelling gelatin derivatives are suitable for special 3D structuring methods such as inkjet-printing and two photon polymerization.

Packaging

1 g in bottle

Legal Information

X-Pure is a registered trademark of Rousselot B.V. LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Rebers et al.
Gels (Basel, Switzerland), 5(1) (2019-01-27)
Chemically cross-linkable gelatin methacryloyl (GM) derivatives are getting increasing attention regarding biomedical applications. Thus, thorough investigations are needed to achieve full understanding and control of the physico-chemical behavior of these promising biomaterials. We previously introduced gelatin methacryloyl acetyl (GMA) derivatives
Shoko Yamamoto et al.
Journal of materials science. Materials in medicine, 28(3), 48-48 (2017-02-09)
Calcium phosphate cements (CPCs), consisting of a mixture of calcium phosphate powders and setting liquid, have been widely used in orthopedic applications. One of the drawbacks of CPCs is their poor resorbability in the living body, which hinders substitution with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service