Skip to Content
Merck
All Photos(2)

Key Documents

F9130

Sigma-Aldrich

Furaltadone

Synonym(s):

5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4O6
CAS Number:
Molecular Weight:
324.29
Beilstein:
8130725
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(\C=N\N2CC(CN3CCOCC3)OC2=O)o1

InChI

1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2/b14-7+

InChI key

YVQVOQKFMFRVGR-VGOFMYFVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furaltadone is a nitrofuran containing a 5-nitrofuran ring and is a member of synthetic antibiotics.

Application

Furaltadone has been used as a reference standard in liquid chromatography-mass spectrometry (LC-MS/MS) quantifying nitrofuran metabolites in aquaculture matrixes and honey.

Biochem/physiol Actions

Nitrofurans are commercially used as feed additives to promote growth in various fields such as aquaculture, bee colonies, and livestock.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian T Veach et al.
Journal of AOAC International, 101(3), 897-904 (2017-12-01)
This paper describes a rapid and robust method utilizing a single liquid-liquid extraction for the quantitation and confirmation of chloramphenicol, florfenicol, and nitrofuran metabolites in honey. This methodology combines two previous extraction methods into a single extraction procedure and utilizes
Nitrofuran antibiotics: a review on the application, prohibition and residual analysis
Vass M, et al.
Veterinary medicine (Auckland, N.Z.), 53(9), 469-469 (2008)
B Murugasu-Oei et al.
The Journal of antimicrobial chemotherapy, 46(6), 917-919 (2000-12-05)
Depletion of oxygen triggers the shift-down of Mycobacterium bovis BCG to a state of dormancy. Bacilli in their dormant state are resistant to standard anti-mycobacterials. The nitroimidazole metronidazole was the first compound identified to show bactericidal activity against dormant tubercle
Jorge Barbosa et al.
Journal of agricultural and food chemistry, 60(17), 4227-4234 (2012-04-11)
The use of nitrofurans as veterinary drugs has been banned from intensive animal production in the European Union (EU) since 1993. The objective of the present study was to evaluate the accumulation and depletion of furaltadone and nifursol and their
Peng Jie Luo et al.
Biomedical and environmental sciences : BES, 25(4), 449-457 (2012-10-03)
To determine 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ) residues released from protein bound AMOZ in animal tissues. Polyclonal and monoclonal antibodies were produced in this study. A rapid, sensitive, and specific competitive direct enzyme-linked immunosorbent assay (cdELISA) was developed. Rabbit polyclonal antibodies were used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service