Skip to Content
Merck
All Photos(1)

Key Documents

F8053

Sigma-Aldrich

Anti-FADD antibody, Mouse monoclonal

clone FD19, purified from hybridoma cell culture

Synonym(s):

Anti-Fas Associated Death Domain

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FD19, monoclonal

form

buffered aqueous solution

mol wt

antigen ~28 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 0.5-1 μg/mL using total cell extracts of A431 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FADD(8772)

General description

Anti-FADD antibody, Mouse monoclonal (mouse IgG1) is derived from the FD19 hybridoma produced by the fusion of murine myeloma cells (NS1) and splenocytes from mouse immunized with purified recombinant human FADD.

Immunogen

purified recombinant human FADD.

Application

Anti-FADD antibody, Mouse monoclonal has been used in:
  • immunoblotting
  • immunoprecipitation
  • enzyme-linked immunosorbent assay (ELISA)

Biochem/physiol Actions

FADD (Fas Associated Death Domain) is associated with various non-apoptotic cellular pathways such as regulation of cell cycle machinery in T lymphocytes connected to the phosphorylation state of FADD and to the FasL/TRAIL-induced transcriptional activation of c-fos protooncogene. It also interacts with the hepatitis C virus core protein in the HEK-293 cell line.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Huang et al.
International journal of oncology, 49(1), 153-163 (2016-05-24)
The discovery of the TRAIL protein and its death receptors DR4/5 changed the horizon of cancer research because TRAIL specifically kills cancer cells. However, the validity of TRAIL-based cancer therapies has yet to be established, as most cancer cells are
Qinghong Kong et al.
International journal of molecular sciences, 19(10) (2018-10-03)
Naphthyridine derivatives are a widely-used class of heterocycles due to their pharmacological activities. A novel compound (10-Methoxy-1,2,3,4-tetrahydrobenzo(g)(1,3) diazepino(1,2-a)-(1,8)naphthyridin-6-yl)(phenyl) methanone (named 3u), showed good anticancer activity in the human malignant melanoma cell line A375 via Thiazolyl Blue Tetrazolium Bromide (MTT) assay.
A novel naphthyridine derivative, 3u, induces necroptosis at low concentrations and apoptosis at high concentrations in human melanoma A375 cells
Kong Q, et al.
International Journal of Molecular Sciences, 19(10), 2975-2975 (2018)
Overcoming resistance to TRAIL-induced apoptosis in solid tumor cells by simultaneously targeting death receptors, c-FLIP and IAPs
Huang Y, et al.
International Journal of Oncology, 49(1), 153-163 (2016)
Apo2L/TRAIL and the death receptor 5 agonist antibody AMG 655 cooperate to promote receptor clustering and antitumor activity
Graves JD, et al.
Cancer Cell, 26(2), 177-189 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service