Skip to Content
Merck
All Photos(1)

Documents

F6162

Sigma-Aldrich

Anti-Fibroblast Growth Factor-Basic antibody, Mouse monoclonal

clone FB-8, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Fibroblast Growth Factor-Basic antibody produced in mouse, Anti-FGF2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FB-8, monoclonal

form

buffered aqueous solution

mol wt

antigen 18 kDa

species reactivity

bovine, human

technique(s)

dot blot: suitable
indirect ELISA: suitable
neutralization: 5 μg/mL using 1 unit recombinant human FGF2 in mouse 3T3 cells
radioimmunoassay: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FGF2(2247)

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Fibroblast Growth Factor-Basic antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the FB-8 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Fibroblast growth factors (FGFs) are members of a family of polypeptides. The names FGF acidic and basic are used for FGF-1 and FGF-2, respectively. Acidic and basic fibroblast growth factor (aFGF and bFGF, respectively) are members of a small family of heparin binding growth factors (HBGF). bFGF usually has an apparent mass of about 18 kDa.

Specificity

The antibody reacts specifically with both native and recombinant human FGF2. It does not cross-react with human FGF1.

Immunogen

recombinant human FGF2.

Application

Anti-Fibroblast Growth Factor-Basic antibody has been used in in immunolabelling and western blotting. It may also be used in:
  • in immunoblotting
  • in enzyme linked immunosorbent assay (ELISA)
  • in dot blot
  • in radio immune assay (RIA)
  • in neutralization bioassays

Used in protein pin array assay to identify the interactive sequences in human αB crystalline protein. The antibody can be used for neutralization of the biological activity of FGF2 in cell culture.

Biochem/physiol Actions

Fibroblast Growth Factor Basic (also known as bFGF) is a pleotropic cytokine that that is a potent mitogenic factor. On binding to its cognate receptor tyrosine kinase, FGFR, bFGF is capable of activating pathways such as MAPK and PI3K and produce cellular effects in varied cell types. Fibroblast Growth Factor-basic is reported to induce proliferation of fibroblasts, differentiation, wound healing and regeneration, limb formation, tissue remodeling and angiogenesis.

Physical form

Solution, 0.2 μm filtered, in phosphate buffered saline, pH 7.4.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Sulpice et al.
Blood, 100(9), 3087-3094 (2002-10-18)
Platelet factor 4 (PF-4) is a member of the chemokine family with powerful antiangiogenic properties. The mechanism by which PF-4 inhibits endothelial cell proliferation is unclear. We investigated the effects of PF-4 on the intracellular signal transduction induced by basic
M Arras et al.
The Journal of clinical investigation, 101(1), 40-50 (1998-02-14)
We have previously shown that monocytes adhere to the vascular wall during collateral vessel growth (arteriogenesis) and capillary sprouting (angiogenesis). In this study we investigated the association of monocyte accumulation with both the production of the cytokines-basic fibroblast growth factor
Ultrastructural immunolocalization of basic fibroblast growth factor in fibroblasts and extracellular matrix
Aktas RG and Kayton RJ
Histochemistry and Cell Biology, 113(3), 227-233 (2000)
S A Boldin et al.
The Journal of biological chemistry, 275(14), 9905-9909 (2000-04-01)
We have previously shown that ongoing glucosylceramide (GlcCer) synthesis is required for basic fibroblast growth factor (bFGF) and laminin to stimulate axonal growth in cultured hippocampal neurons (Boldin, S., and Futerman, A. H. (1997) J. Neurochem. 68, 882-885). We now
B P Mari et al.
The Journal of biological chemistry, 273(1), 618-626 (1998-02-07)
Stromelysin-3 (STR-3) is a recently characterized matrix metalloproteinase (MMP) with a unique pattern of expression and substrate specificity. Unlike other MMPs, STR-3 is consistently and dramatically overexpressed by multiple epithelial malignancies, including carcinomas of the breast, lung, colon, head and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service