Skip to Content
Merck
All Photos(1)

Key Documents

F7807

Sigma-Aldrich

Prostaglandin F Synthase human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

17-beta-hydroxysteroid dehydrogenase type 5, AKR1C3, aldo-keto reductase family 1 member C3, indanol dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

description

contains a C-terminal hexahistidine tag

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥0.4 μmol/mg protein

mol wt

~37 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AKR1C3(8644)

Application

Prostaglandin F synthase (PGF synthase) is a member of the aldo-keto reductase superfamily of proteins which catalyze the conversion of aldehydes and ketones to thier corresponding alcohols. Prostaglandin F synthase is used in cancer research since it is upregulated in many cancers such as breast and prostate cancer. PGF synthase is also used to study kidney and renal cell carcinoma .

Biochem/physiol Actions

PGF synthase catalyzes the conversions of 4-androstene-3,17-dione to testosterone, estrone to 17β-estradiol, prostaglandin H2 to PGF2α, and PGD2 to 9α,11β-PGF2. Prostaglandin (PG) F2 is synthesized via three pathways .

Unit Definition

One unit is defined as the amount of enzyme required to produce 1 μmol of NADP per minute at 37°C in 50 mM KPO4 pH 7.2 containing 250 μM NADPH and 25 μM 9,10-phenanthrenequinone

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal H Wrobel et al.
Toxicology, 262(3), 224-229 (2009-06-25)
Polychlorinated biphenyls (PCBs) stimulate in vitro both the force of myometrial contractions and endometrial secretion of PGF2alpha in cattle. Therefore, the goal of this study was to investigate the participation of PGF2alpha in the effect of PCBs on uterine contractility.
Agnieszka Waclawik et al.
Reproduction (Cambridge, England), 140(4), 613-622 (2010-07-10)
Oxytocin (OXT) and tumor necrosis factor α (TNF) have been implicated in the control of luteolysis by stimulating endometrial secretion of luteolytic prostaglandin F(2α) (PGF(2α)). Nevertheless, OXT concentration in porcine uterine lumen increases markedly on days 11-12 of pregnancy, and
Trevor M Penning et al.
Annals of the New York Academy of Sciences, 1155, 33-42 (2009-03-03)
Prostate and breast cancer are hormone-dependent malignancies of the aging male and female and require the local production of androgens and estrogens to stimulate cell proliferation. Aldo-keto reductases (AKR) play key roles in this process. In the prostate, AKR1C3 (type
Dominika Slonina et al.
Prostaglandins & other lipid mediators, 90(3-4), 69-75 (2009-09-15)
The aim was to study whether bovine myometrial cells have an enzymatic system able to produce prostaglandins (PGs) and whether PGs synthesis is regulated by steroids in a similar manner as endometrial cells. In Experiment 1, immunohistochemical studies localized proteins
Marta Wasielak et al.
Reproduction, fertility, and development, 21(5), 709-717 (2009-06-03)
The present study was conducted to evaluate the effect of the conceptus on uterine prostaglandin-F2alpha (PGF2alpha) and prostaglandin-E2 (PGE2) release and the expression of prostaglandin synthase enzymes during the periimplantation period in the pig. A surgically generated model with conceptuses

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service