Skip to Content
Merck
All Photos(2)

Key Documents

RAB0411

Sigma-Aldrich

Human CEA ELISA Kit

for serum, plasma, cell culture supernatant and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 200 pg/mL
standard curve range: 0.343-250 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PSG2(5670)

General description

The Human CEA (Carcinoembryonic Antigen) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CEA in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human CEA

Application

Human CEA ELISA Kit has been used to compare the analytical results of paper-based analytical device (PAD)-based sensing platform with commercial enzyme linked immunosorbent asay (ELISA) kits for measuring target carcinoembryonic antigen (CEA) levels. It has also been used to comparatively study the limit of detection (LOD) of the quartz crystal microbalance (QCM) immunosensing method.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenli Qiu et al.
Analytical chemistry, 89(9), 5152-5160 (2017-04-06)
An all-in-one paper-based analytical device (PAD) was successfully developed for visual fluorescence detection of carcinoembryonic antigen (CEA) on CdTe/CdSe quantum dot (QD)-enzyme-impregnated paper by coupling with a bioresponsive controlled-release system from DNA-gated mesoporous silica nanocontainers (MSNs). The assay was carried
Enhanced sensitivity of quartz crystal microbalance immunosensor via back-conjugation of biofunctionalized magnetic beads with an external magnetic field
Liao J, et al.
Biochemical Engineering Journal, 114, 276-282 (2016)
Liangjie Chi et al.
The Analyst, 145(18), 6111-6118 (2020-08-26)
Methods based on enzyme labels or nano labels have been developed for immunoassays, but most of them have low sensitivity and are unsuitable for low-abundance protein diagnostics and biosecurity. Herein, an innovative quartz crystal microbalance (QCM) immunosensing method was designed
Hai-Feng Yun et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 3896948-3896948 (2020-08-25)
Ulcerative colitis (UC) is a chronic recurrent inflammation of the colon, and clinical outcome of UC is still unsatisfied. Pingkui enema, a traditional Chinese medicine prescription, has been safely applied for the treatment of diarrhea and dysentery in clinic for
Melvin E Klegerman et al.
Journal of liposome research, 24(3), 216-222 (2014-03-07)
Thermodynamic analysis of ligand-target binding has been a useful tool for dissecting the nature of the binding mechanism and, therefore, potentially can provide valuable information regarding the utility of targeted formulations. Based on a consistent coupling of antibody-antigen binding and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service