Skip to Content
Merck

Skip To

F7250

Fluorescein isothiocyanate isomer I

≥90% purity (HPLC), powder

Synonym(s):

FITC, Fluorescein 5-isothiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H11NO5S
CAS Number:
Molecular Weight:
389.38
NACRES:
NA.47
PubChem Substance ID:
UNSPSC Code:
12171500
EC Number:
222-042-0
MDL number:
Beilstein/REAXYS Number:
1407295
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder

Quality Segment

assay

≥90% (HPLC)

form

powder

technique(s)

titration: suitable

color

orange to dark orange

mp

>360 °C (lit.)

solubility

acetone: 1 mg/mL

fluorescence

λex 492 nm; λem 518 nm (green)

suitability

suitable for protein labeling

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)N=C=S)c1

InChI

1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H

InChI key

MHMNJMPURVTYEJ-UHFFFAOYSA-N

General description

Fluorescein isothiocyanate (FITC) is yellow-orange in color with an absorption maximum at 495nm. Upon excitation, it emits a yellow-green color with an emission maximum at 525nm.
It is widely used to attach a fluorescent label to proteins via the amine group. The isothiocyanate group reacts with amino terminal and primary amines in proteins. It has been used for the labeling of proteins including antibodies and lectins.
Fluorescein isothiocyanate isomer I has been proposed as a contact sensitizer.

Application

Reagent for the FITC labeling of proteins; Microsequencing of proteins and peptides (HPLC)
Reagent for the FITC labeling of proteins; microsequencing of proteins and peptides (HPLC)
Biological applications include use as a fluorescent labeling reagent for proteins, a fluorescent reagent for protein tracing, and a reagent in the fluorescent antibody technique for the rapid identification of pathogens. It may be employed as the derivatization reagent for amphetamine, methamphetamine, 3,4-methylenedioxymethamphetamine and P-phenylethylamine in human urine during their capillary electrophoretic (CE) determination. It may be used for the preparation of fluorescent antibodies. It was employed for in vitro sensitization studies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F427446951F3651
description

suitable for protein labeling, ≥90% (HPLC), powder

description

≥97.5% (HPLC)

description

≥97.5% (HPLC)

description

≥90% (HPLC)

technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

-

technique(s)

titration: suitable

suitability

suitable for protein labeling

suitability

-

suitability

-

suitability

-

color

orange to dark orange

color

yellow to very dark yellow

color

-

color

yellow to orange

fluorescence

λex 492 nm; λem 518 nm (green)

fluorescence

λex 492 nm; λem 518 nm (green)

fluorescence

λex 492 nm; λem 518 nm (green), λex 492 nm; λem 518 nm in 0.1 M phosphate pH 8.0

fluorescence

-

solubility

acetone: 1 mg/mL

solubility

acetone: soluble 10 mg/mL

solubility

acetone: soluble 0.1 g/10 mL

solubility

acetone: 1 mg/mL, clear, light yellow to yellow

form

powder

form

powder

form

powder

form

powder


Still not finding the right product?


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–6 of 6 Questions  
  1. Are cell membranes of living cells permeable for FITC?

    1 answer
    1. FITC is not membrane permeable in healthy cells, it binds to surface proteins. Conjugates such as FITC-Dextran is an example of a more cell-permeable option.

      Helpful?

  2. Is the intensity of FITC (e.g., Product No. F7250) pH dependent?

    1 answer
    1. Yes; many articles have reported the pH dependence of the fluorescence of fluorescein derivatives. See the following references: Archives Biochem. Biophys., 133, 263 (1969); J. Immunol., 95, 1165 (1966); Archives Biochem. Biophys., 73, 1 (1958); Standard test for labeling efficiency of FITC with BSA: NCCLS Approved Standard: ASM-1, 2nd ed., June, 1975; Characterization of FITC-dyes by IR and TLC: Analytical Biochem., 29, 526 (1969); Fluorescent labeling of proteins: Proc. Nat. Acad. Sci.,  72, 459 (1975); Amer. J. Pathol., 34, 1081 (1958); Fluorescent labeling of a lectin: Meth. Enzymol., 81, 65 (1982).

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  4. After the FITC (e.g., Product No. F7250) is conjugated to the protein, what is the solution stability of the conjugated protein?

    1 answer
    1. The stability of the conjugated protein will be essentially the stability of the unconjugated protein.  Some conjugated proteins might be stable when stored at -20°C and other conjugated proteins will have better solution stability at 2-8°C  If the protein concentration is too low, this also has a negative effect upon the stability of the conjugated protein.

      Helpful?

  5. Is Product No. F7250, Fluorescein isothiocyanate isomer I, pharmaceutical grade?

    1 answer
    1. No. The product is considered suitable for research use only.

      Helpful?

  6. How stable is FITC Isomer 1 (e.g., Product No. F7250) once conjugated to an antibody?

    1 answer
    1. FITC can photo bleach fairly rapidly. The prepared solution should be stored in a dark container and protected from light. When viewing the antibody staining under a microscope, it is advisable to use subdued fluorescent light when possible. If the fluorescent light stays at one position for too long, photo bleaching is noticeable.

      Helpful?

Reviews

No rating value

Active Filters