Skip to Content
Merck
All Photos(4)

Key Documents

F3393

Sigma-Aldrich

Anti-Fibroblast Growth Factor-Basic (1-24) antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti-FGF2, Anti-bFGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, bovine, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1:1,000

impurities

endotoxin, tested

UniProt accession no.

application(s)

research pathology

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

bovine ... FGF2(281161)
human ... FGF2(2247)
rat ... Fgf2(54250)

General description

Fibroblast growth factor-basic (FGFb), also known as NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6) is encoded by the gene mapped to human chromosome 4q28.1. The FGFb gene spanning 34271 bp on genomic DNA contains 16 exons and six introns. FGFb RNA transcripts are expressed in various human tissues including liver, thymus, spleen, peripheral blood leukocytes, heart skeletal muscle, testis, colon and kidney.

Immunogen

synthetic peptide sequence corresponding to bovine FGF2 (1-24).

Application

Anti-Fibroblast Growth Factor-Basic (1-24) antibody produced in rabbit has been used in:
  • immunohistochemistry
  • immunostaining
  • immunoblotting

Biochem/physiol Actions

Fibroblast Growth Factor Basic (also known as bFGF) is a pleotropic cytokine that that is a potent mitogenic factor. On binding to its cognate receptor tyrosine kinase, FGFR, bFGF is capable of activation of pathways such as MAPK and PI3K and produce cellular effects in varied cell types. Fibroblast Growth Factor-basic is reported to induce proliferation of fibroblasts, differentiation, wound healing and regeneration, limb formation, tissue remodeling and angiogenesis.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

Bioactivity: Anti-Fibroblast Growth Factor-Basic (1-24) caused a dose-dependent decrease in the mitogenic activity of FGF-Basic such that a final dilution of 1:50 neutralized the bioactivity of 1 unit of FGF Basic.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Qian Liu et al.
Experimental and therapeutic medicine, 23(3), 216-216 (2022-02-08)
Hypertension is associated with bladder symptoms. The present study investigated whether an angiotensin receptor blocker could improve the symptoms and pathological changes associated with a neurogenic bladder (NB). A Sprague-Dawley rat model of NB was constructed. Rats in the sham
Effect of human umbilical cord blood mesenchymal stem cells administered by intravenous or intravitreal routes on cryo-induced retinal injury
Mohamed EM, et al.
IUBMB Life, 69(3), 188-201 (2017)
Correlation of astrocyte elevated gene-1, basic-fibroblast growth factor, beta-catenin, Ki-67, tumor necrosis factor-alfa with prognostic parameters in ductal carcinomas and ductal intraepithelial neoplasms
Erdem H, et al.
Nigerian Journal of Clinical Practice, 18(5), 638-643 (2015)
B G Condron
Neuron, 24(3), 531-540 (1999-12-14)
In the grasshopper CNS, serotonergic growth cones cross the midline early in development and initiate expression of serotonin uptake activity, or SERT. To test if the midline contains an activity that induces SERT, cuts were made that separated serotonergic cell
Joost F M Lensen et al.
Journal of the American Society of Nephrology : JASN, 16(5), 1279-1288 (2005-03-25)
Glycosaminoglycans (GAG) play an important role in renal homeostasis. They are strongly negatively charged polysaccharides that bind and modulate a myriad of proteins, including growth factors, cytokines, and enzymes. With the aid of specific phage display-derived antibodies, the distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service