Skip to Content
Merck
All Photos(1)

Key Documents

F5251

Sigma-Aldrich

p-Fluoro-DL-phenylalanine

≥98.0% (TLC)

Synonym(s):

4-fluoro-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
183.18
Beilstein:
2938793
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

p-Fluoro-DL-phenylalanine,

Assay

≥98.0% (TLC)

form

powder

color

white to off-white

mp

253-255 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NC(Cc1ccc(F)cc1)C(O)=O

InChI

1S/C9H10FNO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)

InChI key

XWHHYOYVRVGJJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits mitosis and reversibly arrests HeLa cells in G2.
p-Fluoro-DL-phenylalanine (pFPhe), a toxic antimetabolite, is a racemic mixture of a substituted (halogenated) benzoyl D and L phenylalanine useful as a selection marker for wine yeast transformation and with potential use in endomorphin, antiviral, and antimicrobial drug development. Phenylalanine analogue, rho-fluorophenylalanine (pFPhe) is also used to induce apoptosis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hae Sun Park et al.
Biochemical and biophysical research communications, 377(1), 280-285 (2008-10-11)
Phenylalanine analog, rho-fluorophenylalanine (pFPhe)-mediated cytotoxicity and several apoptotic events including mitochondrial cytochrome c release, activation of caspase-9, -3, and -8, Bid cleavage, degradation of PARP and PLCgamma-1, and DNA fragmentation were more significant in p56(lck)-deficient Jurkat T cells (JCaM1.6) than
Eduardo Cebollero et al.
Applied and environmental microbiology, 70(12), 7018-7023 (2004-12-03)
Genetic improvement of industrial yeast strains is restricted by the availability of selectable transformation markers. Antibiotic resistance markers have to be avoided for public health reasons, while auxotrophy markers are generally not useful for wine yeast strain transformation because most
H Kloosterman et al.
Microbiology (Reading, England), 149(Pt 11), 3321-3330 (2003-11-06)
Prephenate dehydratase (PDT), chorismate mutase (CM) and 3-deoxy-D-arabino-7-heptulosonate 7-phosphate (DAHP) synthase are key regulatory enzymes in aromatic amino acid biosynthesis in the actinomycete Amycolatopsis methanolica. Deregulated, feedback-control-resistant mutants were isolated by incubation of A. methanolica on glucose mineral agar containing
Jayapal Reddy Mallareddy et al.
Journal of medicinal chemistry, 54(5), 1462-1472 (2011-02-04)
This study reports on new proteolytically stable, pharmacologically active endomorphin analogues, incorporating Dmt(1), Achc(2), pFPhe(4), or βMePhe(4) unnatural amino acids. Consistent with earlier results, it was found that the analogues carrying Dmt(1) and Achc(2) residues displayed the highest μ-opioid receptor
Arthur Hawtrey et al.
Nucleosides, nucleotides & nucleic acids, 27(9), 1011-1023 (2008-08-20)
N-Acylated derivatives of 8-(6-aminohexyl) amino-adenosine-5 '-phosphate were prepared and studied with regard to their effect on DNA synthesis by the Moloney leukemia virus reverse transcriptase. N-palmitoyl and N-nicotinyl derivatives and bis-8-(6-aminohexyl) amino-5'-AMP inhibited the enzyme partially using poly (rA).oligo d(pT)(16-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service