Skip to Content
Merck
All Photos(1)

Key Documents

EP951031607

Eppendorf® Deepwell plates, DNA LoBind, 384 wells

white plate, conical bottom, colorless wells, capacity 200 μL, pack of 120 ea (10 bags × 12 plates)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
material:
clear wells
colorless wells
conical bottom
polypropylene
white plate
size:
384 well
sterility:
non-sterile
binding type:
low binding surface
feature:
lid: no

material

clear wells
colorless wells
conical bottom
polypropylene
white plate

sterility

non-sterile

feature

lid: no

packaging

pack of 120 ea (10 bags × 12 plates)

manufacturer/tradename

Eppendorf® 951031607

capacity

200 μL

size

384 well

well volume

250 μL

color

white border

suitability

suitable for PCR
suitable for molecular biology

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Deepwell Plate 384/200µL, DNA LoBind, wells clear, 200 µL, LoBind, PCR clean, white, 120 plates (10 bags × 12 plates)
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Affinity Purification of Yeast Protein-interacting Metabolites for ESI-MS Analysis
Xiyan Li
Bio-protocol, 1, 45-45 (2011)
Human tear peptide/protein profiling study of ocular surface diseases by SPE-MALDI-TOF mass spectrometry analyses
EuPA open proteomics, 3, 206-215 (2014)
Peptidomics of the agriculturally damaging larval stage of the cabbage root fly Delia radicum (Diptera: Anthomyiidae)
Judith Zoephel
PLoS ONE, 7, 7-7 (2012)
Nikos Tsolakos et al.
Vaccine, 28(18), 3211-3218 (2010-03-02)
In this study, we evaluated the effect of the growth medium on the composition and immunogenicity of meningococcal outer membrane vesicle (OMV) vaccines after cultivation of the Norwegian serogroup B 44/76 vaccine strain in either Frantz' or modified Catlin-6 media
M Yang et al.
International journal of pharmaceutics, 331(2), 176-181 (2006-11-28)
Salmon calcitonin (sCT) powders suitable for inhalation, containing chitosan and mannitol as absorption enhancer and protection agent, respectively, were prepared using a spray-drying process. The effect of chitosan on physicochemical stability of sCT in the dry powder was investigated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service