Merck
  • Characterisation of salmon calcitonin in spray-dried powder for inhalation. Effect of chitosan.
Characterisation of salmon calcitonin in spray-dried powder for inhalation. Effect of chitosan.
International journal of pharmaceutics (2006-11-28)
M Yang, S Velaga, H Yamamoto, H Takeuchi, Y Kawashima, L Hovgaard, M van de Weert, S Frokjaer
ABSTRACT
Salmon calcitonin (sCT) powders suitable for inhalation, containing chitosan and mannitol as absorption enhancer and protection agent, respectively, were prepared using a spray-drying process. The effect of chitosan on physicochemical stability of sCT in the dry powder was investigated by different analytical techniques. High-performance liquid chromatography (HPLC) analysis indicated that sCT was chemically stable upon spray-drying. With the proportion of chitosan in spray-drying formulation being increased, dissolution of sCT from the dry powders was decreased both in phosphate buffer and acetate buffer. The thioflavine T fluorescence assay showed that no fibrils were present in the spray-dried powder. However, sCT partly fibrillated in the phosphate buffer, but not in acetate buffer. Fourier transform infrared (FTIR) spectra showed that the secondary structure of sCT was slightly changed in the dry powder, yet no aggregate signal was observed. Circular dichroism analysis indicated that the structure of sCT in an aqueous formulation was slightly altered by addition of chitosan. Nevertheless, recovery of sCT was not influenced by chitosan in the aqueous formulation as indicated by HPLC analysis. This study suggested that sCT, in absence of any additives, was stable during the spray-drying process under certain conditions. Addition of chitosan affects recovery of sCT from spray-dried powders, which may be due to formation of a partially irreversible complex between the protein and chitosan during the spray-drying process.
Product Description
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Protein LoBind Microplate 384/V-PP, PCR clean, V-bottom clear wells, white border, pack of 240 ea (10 bags × 24 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 120 ea (10 bags × 12 plates)
Eppendorf® DNA LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® DNA LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® DNA LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Protein LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 20 ea (5 bags × 4 plates)