Skip to Content
Merck
All Photos(1)

Key Documents

EHU076751

Sigma-Aldrich

MISSION® esiRNA

targeting human BRD4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAACCCTAACAAGCCCAAGAGGCAGACCAACCAACTGCAATACCTGCTCAGAGTGGTGCTCAAGACACTATGGAAACACCAGTTTGCATGGCCTTTCCAGCAGCCTGTGGATGCCGTCAAGCTGAACCTCCCTGATTACTATAAGATCATTAAAACGCCTATGGATATGGGAACAATAAAGAAGCGCTTGGAAAACAACTATTACTGGAATGCTCAGGAATGTATCCAGGACTTCAACACTATGTTTACAAATTGTTACATCTACAACAAGCCTGGAGATGACATAGTCTTAATGGCAGAAGCTCTGGAAAAGCTCTTCTTGCAAAAAATAAATGAGCTACCCACAGAAGAAACCGAGATCATGATAGTCCAGGCAAAAGGAAGAGGACGTGGGAGGAAAGAAACA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stella Liong et al.
Reproduction (Cambridge, England), 155(6), 573-582 (2018-05-12)
Preeclampsia affects 5% of all pregnancies and is a serious disorder of pregnancy, characterised by high maternal blood pressure, placental hypoxia, fluid retention (oedema) and proteinuria. Women with preeclampsia are associated with exaggerated levels of pro-inflammatory cytokines, chemokines and anti-angiogenic
Sushmita Mustafi et al.
EBioMedicine, 43, 201-210 (2019-04-13)
Bromodomain and extra-terminal inhibitors (BETi) have shown efficacy for the treatment of aggressive triple negative breast cancer (TNBC). However, BETi are plagued by a narrow therapeutic window as manifested by severe toxicities at effective doses. Therefore, it is a limitation
Zeynab Najafova et al.
Nucleic acids research, 45(1), 127-141 (2016-09-22)
Proper temporal epigenetic regulation of gene expression is essential for cell fate determination and tissue development. The Bromodomain-containing Protein-4 (BRD4) was previously shown to control the transcription of defined subsets of genes in various cell systems. In this study we
Xuanchen Zhou et al.
Frontiers in medicine, 7, 413-413 (2020-09-15)
Objectives: This study aimed to explore the relationship between bromodomain-containing protein 4 (BRD4), epithelial-mesenchymal transition (EMT), and disease severity in chronic rhinosinusitis with nasal polyps (CRSwNP). Methods: We performed immunofluorescent (IF) staining to evaluate the expression of BRD4 in the
Hong Zuo et al.
Biochimie, 165, 100-107 (2019-07-22)
High glucose (HG)-induced podocyte injury contributes to the pathogenesis of diabetic nephropathy, a severe complication of diabetes. Bromodomain-containing protein 4 (BRD4) has emerged as a critical regulator for cell injury. However, whether BRD4 participates in HG-induced podocyte injury remains unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service