Skip to Content
Merck
All Photos(1)

Key Documents

EHU024041

Sigma-Aldrich

MISSION® esiRNA

targeting human GATA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGCTTCCTGGAGACTTTGAAGACAGAGCGGCTGAGCCCAGACCTCCTGACCCTGGGACCTGCACTGCCTTCATCACTCCCTGTCCCCAATAGTGCTTATGGGGGCCCTGACTTTTCCAGTACCTTCTTTTCTCCCACCGGGAGCCCCCTCAATTCAGCAGCCTATTCCTCTCCCAAGCTTCGTGGAACTCTCCCCCTGCCTCCCTGTGAGGCCAGGGAGTGTGTGAACTGCGGAGCAACAGCCACTCCACTGTGGCGGAGGGACAGGACAGGCCACTACCTATGCAACGCCTGCGGCCTCTATCACAAGATGAATGGGCAGAACAGGCCCCTCATCCGGCCCAAGAAGCGCCTGATTGTCAGTAAACGGGCAGGTACTCAGTGCACCAACTGCCAGACGACCACCACGACACTGTGGCGGAGAAATGCCAGTGGGGATCCCGTGTGCAATGCCTGCGGCCTCTACTACAAGCTACACCAGGTGAACCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John Timothy Caldwell et al.
PloS one, 8(7), e68601-e68601 (2013-07-23)
It has been previously shown that acute myeloid leukemia (AML) patients with higher levels of GATA1 expression have poorer outcomes. Furthermore, pediatric Down syndrome (DS) patients with acute megakaryocytic leukemia (AMKL), whose blast cells almost universally harbor somatic mutations in
Miranda L Xu et al.
Journal of neurochemistry, 146(4), 390-402 (2018-04-21)
Acetylcholinesterase (AChE; EC 3.1.1.7) is known to hydrolyze acetylcholine at cholinergic synapses. In mammalian erythrocyte, AChE exists as a dimer (G2 ) and is proposed to play role in erythropoiesis. To reveal the regulation of AChE during differentiation of erythroblast
Anouck Wijgaerts et al.
Haematologica, 102(4), 695-706 (2017-01-14)
Gray platelet syndrome is named after the gray appearance of platelets due to the absence of α-granules. It is caused by recessive mutations in
Pavel Burda et al.
PloS one, 11(3), e0152234-e0152234 (2016-03-25)
GATA-1 and PU.1 are two important hematopoietic transcription factors that mutually inhibit each other in progenitor cells to guide entrance into the erythroid or myeloid lineage, respectively. PU.1 controls its own expression during myelopoiesis by binding to the distal URE
A Maroz et al.
Leukemia, 28(6), 1259-1270 (2013-12-18)
Transient leukemia (TL) is evident in 5-10% of all neonates with Down syndrome (DS) and associated with N-terminal truncating GATA1 mutations (GATA1s). Here we report that TL-cell clones generate abundant eosinophils in a substantial fraction of patients. Sorted eosinophils from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service