Skip to Content
Merck
All Photos(1)

Key Documents

P3490

Sigma-Aldrich

Ponasterone A

≥65%

Synonym(s):

2β,3β,14α,20R,22R-Pentahydroxy-5β-cholest-7-en-6-one, 25-Deoxy-20-hydroxyecdysone, 25-Deoxyecdysterone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O6
CAS Number:
Molecular Weight:
464.63
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

form

powder

concentration

≥65%

solubility

ethanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(C)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H44O6/c1-15(2)6-7-23(31)26(5,32)22-9-11-27(33)17-12-19(28)18-13-20(29)21(30)14-24(18,3)16(17)8-10-25(22,27)4/h12,15-16,18,20-23,29-33H,6-11,13-14H2,1-5H3/t16-,18-,20+,21-,22-,23+,24+,25+,26+,27+/m0/s1

InChI key

PJYYBCXMCWDUAZ-JJJZTNILSA-N

Application

Ponasterone A has been used to induce the expression of human huntingtin (HTT).
Suitable as an inducer of ecdysone-inducible mammalian expression system.

Biochem/physiol Actions

Ponasterone A is an analogue of ecdysone.
Ponasterone A is an insect hormone, involved in regulating metamorphosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sem J Aronson et al.
Human gene therapy, 30(10), 1297-1305 (2019-09-11)
Adeno-associated virus (AAV) vector-mediated gene therapy is currently evaluated as a potential treatment for Crigler-Najjar syndrome (CN) (NCT03466463). Pre-existing immunity to AAV is known to hinder gene transfer efficacy, restricting enrollment of seropositive subjects in ongoing clinical trials. We assessed
Daniel L Kiss et al.
Molecular cancer, 16(1), 179-179 (2017-12-29)
FHIT is a genome caretaker/tumor suppressor that is silenced in >50% of cancers. Although it was identified more than 20 years ago, questions remain as to how FHIT loss contributes to cancer, and conversely, how FHIT acts to maintain genome integrity
Hanife Esengil et al.
Nature chemical biology, 3(3), 154-155 (2007-01-24)
The zebrafish has emerged as a versatile model organism for biomedical research, yet its potential has been limited by a lack of conditional reverse-genetic tools. Here we report a chemically inducible gene expression technology that has orthogonality to vertebrate signaling
Toshiyuki Harada et al.
Journal of chemical information and modeling, 51(2), 296-305 (2011-02-01)
Insect growth is regulated by the orchestrated event of ecdysteroids and their receptor proteins. Agonists/antagonists of ecdysteroid receptor are predicted to disrupt normal growth, providing good candidates of new insecticides. A database of over 2 million compounds was subjected to
Donya Tohidi-Esfahani et al.
Pest management science, 67(11), 1457-1467 (2011-05-20)
Certain bisacylhydrazine compounds such as tebufenozide (RH5992) have been shown to act as order-specific insecticides. Their compatibility with predatory Heteroptera, which are used as biological control agents, has also been demonstrated. However, the molecular mode of action of these ecdysone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service