Skip to Content
Merck
All Photos(4)

Key Documents

E0276

Sigma-Aldrich

EPPS

BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)

Synonym(s):

4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulfonic acid), HEPPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20N2O4S
CAS Number:
Molecular Weight:
252.33
Beilstein:
3957385
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified

Assay

≥99.5% (titration)

form

crystalline powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin and Total Aerobic Microbial Count, tested

pH

7.3-8.7

useful pH range

7.3-8.7

pKa (25 °C)

8.0

pKa 

8

mp

237-239 °C (lit.)

solubility

H2O: 25 g + 50 mL, clear, colorless

cation traces

heavy metals (as Pb): ≤5 ppm

λmax

A290 at ≤0.1 (C=33% w/w)

application(s)

diagnostic assay manufacturing

SMILES string

OCCN1CCN(CCCS(O)(=O)=O)CC1

InChI

1S/C9H20N2O4S/c12-8-7-11-5-3-10(4-6-11)2-1-9-16(13,14)15/h12H,1-9H2,(H,13,14,15)

InChI key

OWXMKDGYPWMGEB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EPPS or 4-(2-hydroxyethyl)piperazine-1-propanesulfonic acid (HEPPS) is a zwitterionic buffer substance from the group of good buffers. It effectively maintains constant pH in a solution, which is necessary for many biological and biochemical reactions. EPPS buffer is widely used in cell culture experiments and biological research.

Application

EPPS may be used as a buffer solution to stabilize the pH in sea water samples during the titration experiments.

Features and Benefits

  • BioPerformance Certified, high-quality level (400), ensures reliable and consistent results.
  • Suitable for mammalian cell culture techniques

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The characteristics of Fe speciation and Fe-binding ligands in the Mariana back-arc hydrothermal plumes
Wang H R, et al.
Geochimica et Cosmochimica Acta, 292, 24-36 (2021)
lonization Enthalpies of Some Common Zvvitterionic Hydrogen-lon Buffers (HEPES, PIPES, HEPPS and BES) for Biological Research
Roig T, et al.
Journal of Separation Science, 993(47), 899-90j-899-90j (1993)
Equilibrium studies, synthesis, and characterization of zwitterionic buffer (HEPES and HEPPS) dimethyltin (IV) complexes
Mahmoud M, et al.
Main Group Chemistry, 6(2) (2007)
Feng Xie et al.
Cancer letters, 364(2), 106-117 (2015-05-17)
Cervical cancer is often associated with eosinophil (EOS) infiltration, but the source and the role of EOS are still largely unknown. Our previous work has established that thymic stromal lymphopoietin (TSLP) can stimulate the growth of cervical cancer cell in
Xiannu Jin et al.
Journal of pharmacological and toxicological methods, 70(2), 188-194 (2014-08-26)
Malaria is a major health concern and affects over 300million people a year. Accordingly, there is an urgent need for new efficacious anti-malarial drugs. A major challenge in developing new anti-malarial drugs is to design active molecules that have preferable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service