Skip to Content
Merck
All Photos(1)

Documents

D6140

Sigma-Aldrich

Demeclocycline hydrochloride

powder or crystals, suitable for cell culture

Synonym(s):

7-Chloro-6-demethyltetracycline hydrochloride, Demethylchlorotetracycline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClN2O8 · HCl
CAS Number:
Molecular Weight:
501.31
Beilstein:
3849198
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

product name

Demeclocycline hydrochloride, powder or crystals

Assay

89.5-102.0% anhydrous basis

Quality Level

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

solubility

soluble (1 mg/mL of Demeclocycline Hydrochloride in 0.01 N hydrochloric acid)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

Cl[H].[H][C@]12C[C@@]3([H])[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C1C(=O)c4c(O)ccc(Cl)c4[C@H]2O

InChI

1S/C21H21ClN2O8.ClH/c1-24(2)14-7-5-6-10(16(27)12-9(25)4-3-8(22)11(12)15(6)26)18(29)21(7,32)19(30)13(17(14)28)20(23)31;/h3-4,6-7,14-15,25-26,28-29,32H,5H2,1-2H3,(H2,23,31);1H/t6-,7-,14-,15-,21-;/m0./s1

InChI key

GVSJQNRGSCOSNJ-KBHRXELFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Biochem/physiol Actions

Because it is excreted more slowly than tetracycline, it maintains effective blood levels for longer periods of time.
Demeclocycline hydrochloride is a tetracycline antibiotic derived from a strain of Streptomyces aureofaciens. DMC protects neurons from glutamate induced toxicity by suppressing calpain I and II activities.
Used to study mechanisms of bacterial protein synthesis inhibition at the level of the 30S subunit and aminoacy-tRNA A-site binding.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sierra H Root et al.
Stem cells (Dayton, Ohio), 38(6), 769-781 (2020-02-14)
Bone remodeling and regeneration are dependent on resident stem/progenitor cells with the ability to replenish mature osteoblasts and repair the skeleton. Using lineage tracing approaches, we identified a population of Dmp1+ cells that reside within cortical bone and are distinct
B Athanassiadis et al.
Australian dental journal, 55(2), 150-155 (2010-07-08)
The in vitro antimicrobial activity of a series of endodontic medicaments and their bases against biofilms of Enterococcus faecalis was investigated. The medicaments tested were Pulpdent paste, Ledermix paste, a 50:50 Ledermix and Pulpdent mixture, and a replica of Ledermix
Gabrielle E Foxa et al.
Methods in molecular biology (Clifton, N.J.), 2221, 165-191 (2020-09-27)
Our laboratories have used genetically engineered mouse models (GEMMs) to assess genetic contributions to skeletal diseases such as osteoporosis and osteoarthritis. Studies on the genetic contributions to OA are often done by assessing how GEMMs respond to surgical methods that
Peter F Day et al.
Journal of endodontics, 37(8), 1052-1057 (2011-07-19)
There is evidence to suggest that Ledermix, placed as an intervisit root canal dressing, might improve periodontal healing after replantation of avulsed teeth. As a part of a multicenter randomized controlled trial, we aimed to compare the effect of 2
P Lenherr et al.
International endodontic journal, 45(10), 942-949 (2012-04-18)
To investigate the discoloration potential of endodontic materials using a bovine tooth model. Two hundred and 10 dentine-enamel cuboid blocks (10 × 10 × 3.5 mm) were prepared out of the middle thirds of bovine tooth crowns. Standardized cavities were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service