Skip to Content
Merck
All Photos(1)

Key Documents

O0500000

Oxytetracycline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Hydroxytetracycline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9 · HCl
CAS Number:
Molecular Weight:
496.89
Beilstein:
3853107
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxytetracycline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O9.ClH/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H/t12-,13-,14+,17+,21-,22+;/m1./s1

InChI key

UBDNTYUBJLXUNN-IFLJXUKPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxytetracycline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Getu Kahsay et al.
Journal of pharmaceutical and biomedical analysis, 75, 199-206 (2013-01-02)
A simple, robust and fast high-performance liquid chromatographic method is described for the analysis of oxytetracycline and its related impurities. The principal peak and impurities are all baseline separated in 20 min using an Inertsil C₈ (150 mm × 4.6
Miaomiao Liu et al.
Applied microbiology and biotechnology, 97(19), 8805-8812 (2012-11-29)
To investigate how the microbial community in activated sludge responded to high antibiotic levels, a bench-scale aerobic wastewater treatment system was used to treat oxytetracycline (OTC) mother liquor (OTC-ML). Removal efficiency of chemical oxygen demand decreased from 64.9 to 51.0 %
M Gatti et al.
Theriogenology, 78(8), 1796-1799 (2012-08-29)
Vaginal secretions are an important source of chemical signals, which affect ewes' attractiveness. Moreover, alterations of vaginal flora reduce sexual attractiveness of estrous ewes. As intravaginal sponges containing progestagens (widely used for estrous synchronization) affect vaginal flora, our aims were
A L Stanton et al.
Journal of dairy science, 95(9), 4950-4960 (2012-08-25)
Bovine respiratory disease complex (BRD) is a common disease in weaned dairy calves that incurs economic and welfare costs. This study was an extension of a randomized clinical trial in which a single injection of tulathromycin (TUL) or oxytetracycline (TET)
Guang-Feng Yang et al.
Bioresource technology, 129, 65-71 (2012-12-13)
The short- and long-term effects of oxytetracycline (OTC) on the anaerobic ammonium oxidation (Anammox) process were evaluated. The OTC inhibition of Anammox was substrate-, and especially nitrite-, dependent. The IC50 of OTC in the batch tests on an Anammox mixed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service