Skip to Content
Merck
All Photos(3)

Documents

D5172

Sigma-Aldrich

n-Dodecyl β-D-maltoside

BioXtra, ≥98% (GC)

Synonym(s):

DDM, Lauryl-β-D-maltoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46O11
CAS Number:
Molecular Weight:
510.62
Beilstein:
55318
MDL number:
UNSPSC Code:
12161900
eCl@ss:
32190102
PubChem Substance ID:
NACRES:
NA.25

description

non-ionic

Quality Level

product line

BioXtra

Assay

≥98% (GC)

form

powder

mol wt

micellar avg mol wt 50,000

aggregation number

98

technique(s)

protein purification: suitable
protein quantification: suitable

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

CMC

0.15 mM (20-25°C)

mp

224-226 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C24H46O11/c1-2-3-4-5-6-7-8-9-10-11-12-32-23-21(31)19(29)22(16(14-26)34-23)35-24-20(30)18(28)17(27)15(13-25)33-24/h15-31H,2-14H2,1H3/t15-,16-,17-,18+,19-,20-,21-,22-,23-,24-/m1/s1

InChI key

NLEBIOOXCVAHBD-QKMCSOCLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Dodecyl β-D-maltoside is a non-ionic surfactant belonging to the alkyl polyglucoside family.

Application

Non-ionic detergent for the stabilization and activation of enzymes and for membrane research.
n-Dodecyl β-D-maltoside has been used in a study to assess the effect of detergents on the thermal behavior of elastin-like polypeptides (ELP). It has also been used in a study to investigate how membrane structures will respond to detergent solubilization from amino acid sequences.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rotational Diffusion of Coumarin 153 in Nanoscopic Micellar Environments of n-dodecyl-β-D-maltoside and n-dodecyl-hexaethylene-glycol Mixtures
Hierrezuelo, H. and C. Ruiz
The Journal of Physical Chemistry, 116, 12476-12476 (2012)
Effect of detergents on the thermal behavior of elastin-like polypeptides
Thapa, A.
Biopolymers, 99, 55-55 (2012)
Ryutaro Tokutsu et al.
The Journal of biological chemistry, 287(37), 31574-31581 (2012-07-18)
Photosystem II (PSII) is a multiprotein complex that splits water and initiates electron transfer in photosynthesis. The central part of PSII, the PSII core, is surrounded by light-harvesting complex II proteins (LHCIIs). In higher plants, two or three LHCII trimers
Ka Young Chung et al.
The Journal of biological chemistry, 287(43), 36305-36311 (2012-08-16)
The G protein-coupled β(2)-adrenoreceptor (β(2)AR) signals through the heterotrimeric G proteins G(s) and G(i) and β-arrestin. As such, the energy landscape of β(2)AR-excited state conformers is expected to be complex. Upon tagging Cys-265 of β(2)AR with a trifluoromethyl probe, (19)F
Electrochemistry, AFM, and PM-IRRA spectroscopy of immobilized hydrogenase: role of a hydrophobic helix in enzyme orientation for efficient H2 oxidation.
Alexandre Ciaccafava et al.
Angewandte Chemie (International ed. in English), 51(4), 953-956 (2011-12-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service