Skip to Content
Merck
All Photos(9)

Documents

O8001

Sigma-Aldrich

Octyl β-D-glucopyranoside

≥98% (GC)

Synonym(s):

n-Octyl glucoside, OGP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
Beilstein:
84118
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

Assay

≥98% (GC)

form

powder

mol wt

micellar avg mol wt 25,000

aggregation number

84

CMC

20-25 mM (20-25°C)

transition temp

cloud point >100 °C

storage temp.

−20°C

SMILES string

CCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octyl β --D-glucopyranoside has been used:
  • In varying concentrations to wash Sepharose beads to study the interaction of Albumin-shFcRn in the presence of Octyl β-D-glucopyranoside
  • As a lysis and solubilization solution for bacterial lipoprotein preparation
  • To solubilize the recombinant 47 kDa antigen from E. coli cell
  • As a component of lysis buffer for determination of protein content by BCA assay
  • In deglycolysation process
  • As a component of lysis buffer to prepare mitochondrial extracts
Non-ionic, dialyzable detergent for the solubilization and isolation of membrane proteins. Has been shown to increase the resolution of proteins in 2D gels.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic and physicochemical characterization of the recombinant DNA-derived 47-kilodalton surface immunogen of Treponema pallidum subsp. pallidum.
Chamberlain N R, et al.
Infection and Immunity, 56(1), 71-78 (1988)
Insulin and AMPK regulate fatty acid translocase/CD36 plasma membrane recruitment in cardiomyocytes via RabGAP AS160 and Rab8a Rab GTPase.
Journal of Lipid Research, jlr-M023424 (2012)
Gating of the mitochondrial permeability transition pore by long chain fatty acyl analogs in vivo.
Samovski D, et al.
The Journal of Biological Chemistry, 285(10), 6879-6890 (2010)
The major histocompatibility complex?related Fc receptor for IgG (FcRn) binds albumin and prolongs its lifespan.
The Journal of Experimental Medicine, 197(3), 315-322 (2003)
Ofir Schuster et al.
ACS omega, 6(5), 3525-3534 (2021-02-16)
SARS-CoV-2, the etiologic agent of the COVID-19 pandemic, emerged as the cause of a global crisis. Rapid and reliable clinical diagnosis is essential for effectively controlling transmission. The gold standard assay for SARS-CoV-2 identification is the highly sensitive real-time quantitative

Protocols

This page shows how to solubilize membrane proteins with products from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service