Skip to Content
Merck
All Photos(4)

Key Documents

D2531

Sigma-Aldrich

Dextromethorphan hydrobromide monohydrate

≥99% (TLC)

Synonym(s):

d-3-Methoxy-N-methylmorphinan hydrobromide monohydrate, (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO · HBr · H2O
CAS Number:
Molecular Weight:
370.32
Beilstein:
6453793
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (TLC)

form

powder

SMILES string

Br[H].[H]O[H].[H][C@]12CCCC[C@]13CCN(C)[C@H]2Cc4ccc(OC)cc34

InChI

1S/C18H25NO.BrH.H2O/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;;/h6-7,12,15,17H,3-5,8-11H2,1-2H3;1H;1H2/t15-,17+,18+;;/m1../s1

InChI key

STTADZBLEUMJRG-IKNOHUQMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Dextromethorphan hydrobromide monohydrate has been used as a substrate for cytochrome P450 2D6 enzyme action, in potassium phosphate buffer for measuring P450 activity and is used in enzyme inhibition studies.

Biochem/physiol Actions

Dextromethorphan hydrobromide is an allosteric antagonist at N-methyl D- aspartate (NMDA)-controlled ion channels and antagonist at voltage-dependent channels. Dextromethorphan hydrobromide is useful in treating dry cough.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In Vitro Inhibition and Induction of Human Liver Cytochrome P450 Enzymes by Gentiopicroside: Potent Effect on CYP2A6
Deng Y, et al.
Drug Metabolism and Pharmacokinetics, 28(4), 339-344 (2013)
T Zimmermann et al.
Arzneimittel-Forschung, 45(1), 41-43 (1995-01-01)
The polymorphism of dextromethorphan (CAS 125-71-3) metabolism is dependent on hepatic cytochrom P4502D6 (CYP2D6) activity. The relationship between the CYP2D6 genotype and the dextromethorphan phenotype was studied in 83 healthy unrelated subjects. Genotype was determined by allele-specific polymerase chain reaction
Dependence on dextromethorphan hydrobromide.
Fleming Philip M
British Medical Journal (Clinical Research ed.), 293(6547), 597-597 (1986)
The effect of prototypic sigma ligands on the binding of [3H]dextromethorphan to guinea pig brain.
Klein, et al.
Neuroscience Letters, 91, 175-175 (1989)
J M Musacchio et al.
Life sciences, 45(19), 1721-1732 (1989-01-01)
There is increasing evidence that sigma ligands and dextromethorphan (DM) bind to at least one common high-affinity site. DM and other antitussives do not produce psychotomimetic effects. This suggested that sigma ligands may produce their characteristic effects through another site

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service