E10521
Ethyl 3-aminobenzoate methanesulfonate
98%
Synonym(s):
MS-222, TS 222, Tricaine, Tricaine methanesulfonate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
H2NC6H4CO2C2H5·CH3SO3H
CAS Number:
Molecular Weight:
261.29
Beilstein:
3775022
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
146-149 °C (lit.)
SMILES string
CS(O)(=O)=O.CCOC(=O)c1cccc(N)c1
InChI
1S/C9H11NO2.CH4O3S/c1-2-12-9(11)7-4-3-5-8(10)6-7;1-5(2,3)4/h3-6H,2,10H2,1H3;1H3,(H,2,3,4)
InChI key
FQZJYWMRQDKBQN-UHFFFAOYSA-N
Related Categories
General description
Ethyl 3-aminobenzoate methanesulfonate is mainly used as an anesthetic for fish. It is also used for the euthanasia of all fish species especially zebrafish, which is commonly used in research.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tricaine methane?sulfonate (MS?222) application in fish anaesthesia.
Topic Popovic N
Journal of Applied Ichthyology, 28(4), 553-564 (2012)
Nina De Rocker et al.
Genetics in medicine : official journal of the American College of Medical Genetics, 17(6), 460-466 (2014-09-19)
Submicroscopic deletions of chromosome band 2p25.3 are associated with intellectual disability and/or central obesity. Although MYT1L is believed to be a critical gene responsible for intellectual disability, so far no unequivocal data have confirmed this hypothesis. In this study we
Karen E Sullam et al.
The ISME journal, 9(7), 1508-1522 (2015-01-13)
Diverse microbial consortia profoundly influence animal biology, necessitating an understanding of microbiome variation in studies of animal adaptation. Yet, little is known about such variability among fish, in spite of their importance in aquatic ecosystems. The Trinidadian guppy, Poecilia reticulata
A review of tricaine methanesulfonate for anesthesia of fish.
Carter K
Reviews in Fish Biology and Fisheries, 21(1), 51-59 (2011)
Anesthesia and euthanasia in zebrafish.
Matthews M
ILAR Journal, 53(2), 192-204 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service