Skip to Content
Merck
All Photos(1)

Key Documents

D1530

Sigma-Aldrich

1α,25-Dihydroxyvitamin D3

≥99% (HPLC)

Synonym(s):

1α,25-Dihydroxycholecalciferol, Calcitriol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
Beilstein:
7559394
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058003
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

Quality Level

Assay

≥99% (HPLC)

form

solid

technique(s)

HPLC: suitable

storage temp.

−20°C

SMILES string

[H][C@@]1(CC[C@@]2([H])C(\CCC[C@]12C)=C\C=C3\C[C@@H](O)C[C@H](O)C3=C)[C@H](C)CCCC(C)(C)O

InChI

1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1

InChI key

GMRQFYUYWCNGIN-NKMMMXOESA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Biologically active form of vitamin D3 in calcium absorption and deposition.
Biologically active form of vitamin D3 in calcium absorption and deposition. 1α,25-Dihydroxyvitamin D3 has widespread effects on cellular differentiation and proliferation, and can modulate immune responsiveness, and central nervous system function. Recent studies suggest that 1α,25-dihydroxyvitamin D3 acts as a chemopreventive agent against several malignancies including cancers of the prostate and colon and shows synergy with other anticancer compounds.
Cholecalciferol is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs. 1α-Hydroxyvitamin D3 (alfacalcidol is a synthetic analog that is metabolized to 1,25-dihydroxycholecalciferol, the biologically active form of vitamin D3 in calcium absorption and deposition. 1α,25-Dihydroxyvitamin D3 has widespread effects on cellular differentiation and proliferation, and can modulate immune responsiveness, and central nervous system function. Recent studies suggest that 1α,25-dihydroxyvitamin D3 acts as a chemopreventive agent against several malignancies including cancers of the prostate and colon and shows synergy with other anticancer compounds.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edwin C Y Chow et al.
American journal of physiology. Endocrinology and metabolism, 304(9), E977-E989 (2013-03-14)
The vitamin D receptor (VDR) maintains a balance of plasma calcium and 1α,25-dihydroxyvitamin D3 [1,25(OH)2D3], its natural active ligand, by directly regulating the calcium ion channel (TRPV6) and degradation enzyme (CYP24A1), and indirectly regulating the parathyroid hormone (PTH) for feedback
Joao F de Brito Galvao et al.
Journal of veterinary emergency and critical care (San Antonio, Tex. : 2001), 23(2), 134-162 (2013-04-10)
To review the inter-relationships between calcium, phosphorus, parathyroid hormone (PTH), parent and activated vitamin D metabolites (vitamin D, 25(OH)-vitamin D, 1,25(OH)2 -vitamin D, 24,25(OH)2 -vitamin D), and fibroblast growth factor-23 (FGF-23) during chronic kidney disease (CKD) in dogs and cats.
Ayten Kandilci et al.
Blood, 114(8), 1596-1606 (2009-06-30)
Forced expression of MN1 in primitive mouse hematopoietic cells causes acute myeloid leukemia and impairs all-trans retinoic acid-induced granulocytic differentiation. Here, we studied the effects of MN1 on myeloid differentiation and proliferation using primary human CD34(+) hematopoietic cells, lineage-depleted mouse
Beth J Kirby et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 28(9), 1987-2000 (2013-03-19)
Pregnancy invokes a doubling of intestinal calcium absorption whereas lactation programs skeletal resorption to provide calcium to milk. Postweaning bone formation restores the skeleton's bone mineral content (BMC), but the factors that regulate this are not established. We used Pth-null
Ichiaki Ito et al.
The Journal of clinical investigation, 123(11), 4579-4594 (2013-10-19)
The TGF-β superfamily comprises pleiotropic cytokines that regulate SMAD and non-SMAD signaling. TGF-β-SMAD signal transduction is known to be involved in tissue fibrosis, including renal fibrosis. Here, we found that 1,25-dihydroxyvitamin D3-bound [1,25(OH)2D3-bound] vitamin D receptor (VDR) specifically inhibits TGF-β-SMAD

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service