Skip to Content
Merck
All Photos(1)

Key Documents

CLS3361

Corning® 96 Well EIA/RIA Assay Microplate

flat bottom clear, polystyrene, high binding surface, bag of 20, sterile, lid

Synonym(s):

96 microtiter plate, 96 multiwell plate, 96 well microplate, 96 well plate, EIA/RIA plate, immunoassay plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom clear
polystyrene
polystyrene
round wells
size:
96 wells , 96 well plate
well volume:
330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL
well working volume:
75-200 μL

material

flat bottom clear
polystyrene
polystyrene
round wells

sterility

sterile

feature

lid
skirt
plate format: 96 well standard
well: flat bottom, clear

packaging

case of 100
bag of 20

manufacturer/tradename

Corning 3361

size

96 wells , 96 well plate

well volume

330-360 μL , Note: CLS3366 and CLS3797 have well volume capacities of 330 uL

well working volume

75-200 μL

color

clear plate

suitability

suitable for (EIA/RIA; immunoassays)

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

  • EIA/RIA plates are tested on lot-to-lot basis and the certification results for each lot are made available upon request
  • High binding surface binds medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions
  • Binding capacity of approximately 500 ng of mouse IgG/cm2
  • Ideal for immunoassays
  • Recommended working volume of up to 75 to 200 μL
  • Conforms to standard microplate footprint and dimensions
These 96 well EIA/RIA plates are designed for in vitro diagnostic immunoassays. All EIA/RIA plates are manufactured from special medical-grade polystyrene for uniform binding, high optical clarity and low background absorption. In addition, the Easy-Wash products are part of the Certified Surface Chemistry Programme, which guarantees protein binding performance to be within QA release specifications.

Legal Information

Corning is a registered trademark of Corning, Inc.
Easy-Wash is a trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service