Skip to Content
Merck
All Photos(1)

Key Documents

C9650

Sigma-Aldrich

β-Cyano-L-alanine

≥95%, suitable for ligand binding assays

Synonym(s):

3-cyano-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

β-Cyano-L-alanine, ≥95%

Quality Level

Assay

≥95%

form

powder

technique(s)

ligand binding assay: suitable

color

white

mp

217 °C

storage temp.

−20°C

SMILES string

OC([C@@H](N)CC#N)=O

InChI

1S/C4H6N2O2/c5-2-1-3(6)4(7)8/h3H,1,6H2,(H,7,8)/t3-/m0/s1

InChI key

BXRLWGXPSRYJDZ-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

β-Cyano-L-alanine (BCA) is used as a cystathione γ-lyase (CSE) inhibitor to study the physiological roles of hydrogen sulfide in processes such as angiogenesis and hypertension.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Miura et al.
Journal of biochemistry, 87(5), 1469-1481 (1980-05-01)
The reaction of D-amino acid oxidase [EC 1.4.3.3] (DAO) from porcine kidney with beta-cyano-D-alanine (D-BCNA) was studied. DAO was found to catalyze elimination of the cyano group as well as oxidation of D-BCNA. During the course of the reaction in
Ying-Yuan Pamela Mok et al.
British journal of pharmacology, 143(7), 881-889 (2004-10-27)
Haemorrhagic shock (60 min) in the anaesthetized rat resulted in a prolonged fall in the mean arterial blood pressure (MAP) and heart rate (HR). Pre-treatment (30 min before shock) or post-treatment (60 min after shock) with inhibitors of cystathionine gamma
K Witthohn et al.
Comparative biochemistry and physiology. C, Comparative pharmacology and toxicology, 79(1), 103-106 (1984-01-01)
Beside the known existence of cyanoglucosides (linamarin and lotaustralin) and proteins the neurotoxin beta-cyanoalanine has been demonstrated for the first time in the defensive secretions of animals. It is proposed that beta-cyanoalanine is produced by metabolizing cyanide from the cyanoglucosides.
Tadahiko Masaki et al.
Journal of gastroenterology, 38(1), 37-44 (2003-02-01)
The clinical utility of relative and absolute grading criteria for submucosal invasion in T1 colorectal carcinomas has been controversial. In 51 T1 colorectal carcinomas, depth of submucosal invasion was graded either according to a modified Haggitt's classification (a relative criterion)
Andrew J M Howden et al.
The Plant journal : for cell and molecular biology, 57(2), 243-253 (2008-09-13)
Pseudomonas fluorescens SBW25 is a plant growth-promoting bacterium that efficiently colonizes the leaf surfaces and rhizosphere of a range of plants. Previous studies have identified a putative plant-induced nitrilase gene (pinA) in P. fluorescens SBW25 that is expressed in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service