Skip to Content
Merck
All Photos(1)

Key Documents

81838

Sigma-Aldrich

L-C-Propargylglycine

≥99.0% (TLC)

Synonym(s):

L-Propargylglycine, (S)-2-Amino-4-pentynoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2
CAS Number:
Molecular Weight:
113.11
Beilstein:
2347861
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-C-Propargylglycine, ≥99.0% (TLC)

Assay

≥99.0% (TLC)

form

powder

color

white

mp

235-239 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

N[C@@H](CC#C)C(O)=O

InChI

1S/C5H7NO2/c1-2-3-4(6)5(7)8/h1,4H,3,6H2,(H,7,8)/t4-/m0/s1

InChI key

DGYHPLMPMRKMPD-BYPYZUCNSA-N

Application

Reagent for the irreversible inactivation of γ-cystathionase; affinity labeling reagent for γ-cystathionase and other enzymes; peptides containing this amino acid can be tritiated to high specific radioactivity

Biochem/physiol Actions

L-C-Propargylglycine, a specific inhibitor of H(2)S synthase of cystathionine-γ-lyase (CSE), may be used to study the role of H2S in regulation of biological processes.
L-propargylglycine (PAG), an inhibitor of cystathionine γ-lyase (CSE), is useful in studies of hydrogen sulphide synthesis and bioactivity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bridget Fox et al.
Journal of cellular and molecular medicine, 16(4), 896-910 (2011-06-18)
Hydrogen sulfide (H(2)S) has recently been proposed as an endogenous mediator of inflammation and is present in human synovial fluid. This study determined whether primary human articular chondrocytes (HACs) and mesenchymal progenitor cells (MPCs) could synthesize H(2)S in response to
Heloísa Della Coletta Francescato et al.
Archives of toxicology, 85(12), 1597-1606 (2011-05-19)
A single injection of adriamycin (ADR) induces marked and persistent proteinuria in rats that progress to glomerular and tubulointerstitial lesions. It has been shown that ADR-induced nephrotoxicity is mediated, at least in part, by oxidative stress that lead to inflammation.
Gang Tan et al.
PloS one, 6(10), e25943-e25943 (2011-10-25)
Hydrogen sulfide (H(2)S) displays vasodilative, anti-oxidative, anti-inflammatory and cytoprotective activities. Impaired production of H(2)S contributes to the increased intrahepatic resistance in cirrhotic livers. The study aimed to investigate the roles of H(2)S in carbon tetrachloride (CCl(4))-induced hepatotoxicity, cirrhosis and portal
K Sasaki et al.
Biochemistry international, 10(2), 165-175 (1985-02-01)
Cystathionine accumulated in several tissues of dams and fetuses by a single intraperitoneal administration of L-proparglyglycine to pregnant rats. Cystathionine in the liver of dams reached its maximal level at about 15 hrs after L-proparglyglycine injection (10 mg/300g), while that
Yan Gao et al.
International journal of cardiology, 152(2), 177-183 (2011-02-15)
Hydrogen sulfide (H(2)S) displays anti-inflammatory and cytoprotective activities to attenuate myocardial ischemia-reperfusion (MIR)-induced injury, but its role in MIR in diabetics is not known. This study was undertaken to investigate whether H(2)S plays a protective role in MIR in diabetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service