Skip to Content
Merck
All Photos(3)

Key Documents

C5259

Sigma-Aldrich

Carbamyl-β-methylcholine chloride

≥99% (TLC), crystalline

Synonym(s):

Bethanechol chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H17N2O2Cl
CAS Number:
Molecular Weight:
196.68
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99% (TLC)

form

crystalline

color

white

solubility

H2O: 1.7 g/mL (stable for several days at 4°C.)
ethanol: 80 mg/mL (stable for several days at 4°C.)

storage temp.

2-8°C

SMILES string

Cl.CC(C[N](C)(C)C)OC(N)=O

InChI

1S/C7H17N2O2.ClH/c1-6(11-7(8)10)5-9(2,3)4;/h6H,5H2,1-4H3,(H2,8,10);1H

InChI key

AIRHOJQHGLTTTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Carbamyl-β-methylcholine chloride has been used in western blotting and bactericidal assay. It has also been used in concentration-response studies with isolated atrial preparation.

Biochem/physiol Actions

Carbamyl-β-methylcholine has a selective action on urinary bladder and intestine. It induces smooth muscle tension in the bladder. Its main function is to induce detrusor muscle action, mediated by postganglionic parasympathetic effector cells. Nausea, flushing, sweating, abdominal cramps are some of the side effect caused by carbamyl-β-methylcholine.

Caution

Hygroscopic.

Substrates

Muscarinic acetylcholine receptor agonist that is not a substrate for acetylcholinesterase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Remodeling of cardiac cholinergic innervation and control of heart rate in mice with streptozotocin-induced diabetes
Mabe AM and Hoover DB
Autonomic Neuroscience : Basic & Clinical, 162(1-2), 24-31 (2011)
Pharmacology for the Primary Care Provider, 380-380 (2008)
Darryl J Adamko et al.
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics, 14(5), 321-327 (2012-05-23)
Diagnosing asthma in infancy is largely made on the basis of the symptoms of cough and wheezing. A similar presentation can be seen in neurologically normal infants with excessive gastroesophageal reflux (GER). There are no randomized placebo controlled studies in
Penn Clinical Manual of Urology, 454-454 (2014)
Ana Beatriz Albino de Almeida et al.
Journal of medicinal food, 15(4), 378-383 (2011-12-24)
Arctium lappa L. has been used in folk medicine as a diuretic, depurative, and digestive stimulant and in dermatological conditions. The mechanisms involved in the anti-ulcerogenic activity of the sesquiterpene onopordopicrin (ONP)-enriched fraction (termed the ONP fraction), obtained from A.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service