Skip to Content
Merck
All Photos(1)

Key Documents

C2273

Sigma-Aldrich

Carbonic Anhydrase from bovine erythrocytes

For use as a marker in SDS-PAGE

Synonym(s):

Carbonate Dehydratase, Carbonate Hydrolyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine erythrocytes

Quality Level

form

powder

mol wt

~29 kDa

packaging

vial of 5 mg

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Carbonic Anhydrase is a metalloenzyme with zinc.[1] The active site of carbonic anhydrase contains three conserved histidine residues (His94, His96 and His119).[2]

Application

Carbonic Anhydrase has been used:
  • as a component of nine-protein mixture for the comparative proteomic mass spectrometry analysis of bovine brain samples[3]
  • as a molecular marker in gel filtration chromatography[4]
  • as a standard protein to test the recovery of CMA/20 microdialysis probe[5]

Biochem/physiol Actions

Carbonic anhydrase is essential for the reversible hydration of carbon dioxide.[1] It is also crucial for the physiological processes including respiration, ion transport, acid–base balance, lipid and carbohydrate metabolic pathways.[1] Carbonic anhydrase participates in electrolyte secretion and in the carbon dioxide fixation.[2]

Unit Definition

One collagen digestion unit (CDU) liberates peptides from collagen from bovine achilles tendon equivalent in ninhydrin color to 1.0 μmole of leucine in 5 hours at pH 7.4 at 37 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Imtaiyaz Hassan et al.
Bioorganic & medicinal chemistry, 21(6), 1570-1582 (2012-05-23)
The carbonic anhydrases enzymes (CAs, EC 4.2.1.1) are zinc containing metalloproteins, which efficiently catalyse the reversible conversion of carbon dioxide to bicarbonate and release proton. These enzymes are essentially important for biological system and play several important physiological and patho-physiological
Alan J Rosenbloom et al.
Journal of neuroscience methods, 148(2), 147-153 (2005-07-27)
Unlike all other currently available sampling techniques, microdialysis allows the continuous recovery of dialysed fluid from the interstitial space of living tissue. Microdialysis has been extensively used to study small molecules such as neurotransmitters, metabolites and drugs in the brain
Arun Devabhaktuni et al.
Journal of proteome research, 15(3), 732-742 (2016-01-09)
Dependent on concise, predefined protein sequence databases, traditional search algorithms perform poorly when analyzing mass spectra derived from wholly uncharacterized protein products. Conversely, de novo peptide sequencing algorithms can interpret mass spectra without relying on reference databases. However, such algorithms
Joana da Costa Ores et al.
Chemosphere, 88(2), 255-259 (2012-04-20)
This work presents a study of industrially applicable techniques to obtain a biologically supported carbon dioxide capture system, based on the extraction of carbonic anhydrase from bovine blood. Carbonic anhydrase is a metalloenzyme which catalyzes the reversible hydration of carbon
Fatima Gasset-Rosa et al.
Neuron, 102(2), 339-357 (2019-03-12)
While cytoplasmic aggregation of TDP-43 is a pathological hallmark of amyotrophic lateral sclerosis and frontotemporal dementia, how aggregates form and what drives its nuclear clearance have not been determined. Here we show that TDP-43 at its endogenous level undergoes liquid-liquid

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service