Skip to Content
Merck
All Photos(1)

Key Documents

C2522

Sigma-Aldrich

Carbonic Anhydrase Isozyme II from bovine erythrocytes

lyophilized powder, ≥3,000 W-A units/mg protein

Synonym(s):

Carbonate Dehydratase, Carbonate Hydrolyase, Carbonic Anhydrase II

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine erythrocytes

Quality Level

form

lyophilized powder

specific activity

≥3,000 W-A units/mg protein

mol wt

30 kDa

pI 

~5.9

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Carbonic anhydrase from sigma has been used for the analysis of thermodynamic stability of the enzyme by measuring the binding affinity and stoichiometry.[1] The enzyme has also been used to generate CD4+ T cell lines specific for carbonic anhydrase during the study of autoimmune pancreatitis in rat.[2]

Biochem/physiol Actions

Carbonic anhydrase is a zinc metalloenzyme that has a molecular weight of approximately 30,000 Da. The enzyme catalyzes the hydration of carbon dioxide to carbonic acid. It is involved in many physiological and pathological processes such as pH and CO2 homeostasis, transport of bicarbonate and CO2, biosynthetic reactions, bone resorption, calcification, and tumorigenicity. Therefore, inhibitors that block this enzyme can have therapeutic implications in glaucoma, epilepsy and Parkinson′s disease.[1]

Unit Definition

One Wilbur-Anderson (W-A) unit will cause the pH of a 0.02 M Trizma buffer to drop from 8.3 to 6.3 per min at 0 °C. (One W-A unit is essentially equivalent to one Roughton-Booth unit.)

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gérard Liger-Belair et al.
ACS omega, 8(25), 22844-22853 (2023-07-03)
When it comes to champagne tasting, dissolved CO2 is a key compound responsible for the very much sought-after effervescence in glasses. Nevertheless, the slow decrease of dissolved CO2 during prolonged aging of the most prestigious cuvees raises the issue of
Arne Schön et al.
Analytical biochemistry, 488, 45-50 (2015-08-05)
Protein aggregation is a major issue affecting the long-term stability of protein preparations. Proteins exist in equilibrium between the native and denatured or partially denatured conformations. Often denatured or partially denatured conformations are prone to aggregate because they expose to
Rui Hou et al.
ACS omega, 8(21), 19057-19071 (2023-06-05)
DNA-encoded library (DEL) is a powerful ligand discovery technology that has been widely adopted in the pharmaceutical industry. DEL selections are typically performed with a purified protein target immobilized on a matrix or in solution phase. Recently, DELs have also
Amelia Pollard et al.
Aging, 8(10), 2425-2436 (2016-10-16)
Carbonic anhydrase inhibitors are used to treat glaucoma and cancers. Carbonic anhydrases perform a crucial role in the conversion of carbon dioxide and water into bicarbonate and protons. However, there is little information about carbonic anhydrase isoforms during the process
Hajime Shigemitsu et al.
Nature communications, 11(1), 3859-3859 (2020-08-02)
Non-enzymatic proteins including antibodies function as biomarkers and are used as biopharmaceuticals in several diseases. Protein-responsive soft materials capable of the controlled release of drugs and proteins have potential for use in next-generation diagnosis and therapies. Here, we describe a

Protocols

Objective: To standardize a procedure for the enzymatic assay of Carbonic Anhydrase (EC 4.2.1.1) for Wilbur-Anderson Units.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service