Skip to Content
Merck
All Photos(4)

Key Documents

C1625

Sigma-Aldrich

L-Canavanine

≥98% (TLC), powder, from Canavalia ensiformis

Synonym(s):

L-α-Amino-γ-(guanidinooxy)-n-butyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N4O3
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis

biological source

Canavalia ensiformis

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to yellow

solubility

H2O: <100 mg/mL

application(s)

detection

storage temp.

2-8°C

SMILES string

N[C@@H](CCONC(N)=N)C(O)=O

InChI

1S/C5H12N4O3/c6-3(4(10)11)1-2-12-9-5(7)8/h3H,1-2,6H2,(H,10,11)(H4,7,8,9)/t3-/m0/s1

InChI key

FSBIGDSBMBYOPN-VKHMYHEASA-N

Biochem/physiol Actions

Canavanine is a naturally occurring L-amino acid that interferes with L-arginine-utilizing enzymes due to its structural similarity. It is a selective inhibitor of inducible nitric oxide synthase (iNOS). Overproduction of NO by iNOS plays a crucial role in the pathophysiology of septic shock and chronic inflammation.
A naturally occurring L-amino acid that interferes with L-arginine-utilizing enzymes due to its structural similarity. It is a selective inhibitor of inducible nitric oxide synthase (iNOS). Overproduction of NO by iNOS plays a crucial role in the pathophysiology of septic shock and chronic inflammation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana Konovalova et al.
The international journal of biochemistry & cell biology, 65, 268-274 (2015-06-29)
Impairment of mitochondrial protein homeostasis disrupts mitochondrial function and causes human diseases and aging, but the molecular mechanisms of protein synthesis and quality control in mammalian mitochondria are not fully understood. Here we demonstrate in human cells that misincorporation of
Junjie Tan et al.
Nature communications, 10(1), 439-439 (2019-01-27)
RNA-guided nucleases of the CRISPR/Cas type can be repurposed as programmable nucleotide deaminases to mediate targeted nucleotide substitutions. Such base editors have enormous potential in genome editing, gene therapy and precision breeding. However, current editors suffer from limited specificity in
Margarita Cabrera et al.
The Journal of biological chemistry, 288(7), 5166-5175 (2012-12-25)
Transport along the endolysosomal system requires multiple fusion events at early and late endosomes. Deletion of several endosomal fusion factors, including the Vac1 tether and the Class C core vacuole/endosome tethering (CORVET) complex-specific subunits Vps3 and Vps8, results in a
Karin Lanthaler et al.
BMC biology, 9, 70-70 (2011-10-26)
The uptake of drugs into cells has traditionally been considered to be predominantly via passive diffusion through the bilayer portion of the cell membrane. The recent recognition that drug uptake is mostly carrier-mediated raises the question of which drugs use
Bozhena Vynnytska-Myronovska et al.
International journal of cancer, 130(9), 2164-2175 (2011-06-08)
Single amino acid arginine deprivation is a promising strategy in modern metabolic anticancer therapy. Its potency to inhibit tumor growth warrants the search for rational chemo- and radio-therapeutic approaches to be co-applied. In this report, we evaluated, for the first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service