Skip to Content
Merck
All Photos(1)

Key Documents

H1007

Sigma-Aldrich

L-Homoarginine hydrochloride

≥98% (TLC)

Synonym(s):

(S)-2-Amino-6-guanidinohexanoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NH(CH2)4CH(NH2)CO2H · HCl
CAS Number:
Molecular Weight:
224.69
Beilstein:
4016621
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Homoarginine hydrochloride, unnatural arginine analog

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

213-215 °C (lit.)

solubility

H2O: soluble

application(s)

peptide synthesis

SMILES string

Cl[H].N[C@@H](CCCCNC(N)=N)C(O)=O

InChI

1S/C7H16N4O2.ClH/c8-5(6(12)13)3-1-2-4-11-7(9)10;/h5H,1-4,8H2,(H,12,13)(H4,9,10,11);1H/t5-;/m0./s1

InChI key

YMKBVNVCKUYUDM-JEDNCBNOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Homoarginine may be used to create non-natural proteins for studies of post-translational protein modifications. Arginine residues are often located at the active centers of proteins and enzymes. Replacing these arginines with homoarginine can elucidate protein function and structural requirements. L-Homoarginine is used to study the mechanisms of nitric oxide production by cells via nitric oxide synthase(s). L-Homoarginine is used as a selective mammalian alkaline phosphatase isoenzyme inhibitor. The substitution of homoarginine for arginine or lysine renders proteins resistant to proteolysis by trypsin.
The substitution of homoarginine for arginine or lysine renders proteins resistant to proteolysis by trypsin. Arginine residues are often located at the active centers of proteins and enzymes. Replacing these arginines with homoarginine can elucidate protein function and structural requirements.

Biochem/physiol Actions

Homoarginine is an inhibitor of tissue nonspecific alkaline phosphatase (TNALP). It is also an inhibitor of cellular transport of arginine by a sodium-independent high affinity y+ transporter.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabor Tajti et al.
Allergy, asthma, and clinical immunology : official journal of the Canadian Society of Allergy and Clinical Immunology, 14, 2-2 (2018-01-09)
Contribution of nitric-oxide (NO) pathway to the pathogenesis of bronchial asthma (asthma) is ambiguous as NO may confer both protective and detrimental effects depending on the NO synthase (NOS) isoforms, tissue compartments and underlying pathological conditions (e.g. systemic inflammation). Asymmetric
Roman N Rodionov et al.
Journal of the American Heart Association, 8(14), e012486-e012486 (2019-07-16)
Background Homoarginine ( hA rg) has been shown to be cardioprotective in a model of ischemic heart failure; however, the mechanism remains unknown. hA rg can inhibit tissue-nonspecific alkaline phosphatase ( TNAP ), an enzyme that promotes vascular calcification. We
Michael D Wetzel et al.
Physiological reports, 9(5), e14766-e14766 (2021-03-14)
Recently we showed that homoarginine supplementation confers kidney protection in diabetic mouse models. In this study we tested whether the protective effect of homoarginine is nitric oxide synthase-3 (NOS3)-independent in diabetic nephropathy (DN). Experiments were conducted in NOS3 deficient (NOS3-/-
Amino acids as modulators of endothelium-derived nitric oxide.
Kakoki M, Kim HS, Edgell CJ, et al.
American Journal of Physiology: Renal Physiology, 291, F297-F304 (2006)
Azin Nowrouzi et al.
Biochemical and biophysical research communications, 330(2), 400-409 (2005-03-31)
Liver tissue is the source of 90% of serum alkaline phosphatase (AP). The serum levels and structures of tumor marker proteins change under many disease conditions as well as cancer. The study was aimed at determining the type of alkaline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service